This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs05049715_s1
See other VTI1A GE Assays ›
Gene Symbol:
VTI1A
Gene Name:
vesicle transport through interaction with t-SNAREs 1A
Gene Aliases:
MMDS3, MVti1, VTI1RP2, Vti1-rp2
Chromosome Location:
Chr.10: 112446988 - 112947204 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs05049715_s1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: VTI1A
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_134521.1 4 1537 131
NR_134522.1 3 1533 131
NR_134523.1 2 1425 131
GenBank mRNA AK095476.1 - 4 1529 132

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Gene Ontology Categories:

Function(s) Process(es)

protein targeting to vacuole
ER to Golgi vesicle-mediated transport
intra-Golgi vesicle-mediated transport
Golgi to vacuole transport
autophagy
retrograde transport, endosome to Golgi
vesicle fusion with Golgi apparatus
voluntary musculoskeletal movement
Golgi ribbon formation
SNARE binding
SNAP receptor activity
protein binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1000008927.hg.1 ENST00000393077.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008927.hg.1 ENST00000393077.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008927.hg.1 ENST00000393077.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008927.hg.1 ENST00000393077.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008927.hg.1 ENST00000393077.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008927.hg.1 ENST00000393077.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008927.hg.1 ENST00000393077.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008927.hg.1 ENST00000393077.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008927.hg.1 ENST00000393077.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008927.hg.1 ENST00000393077.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008927.hg.1 ENST00000393077.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008927.hg.1 ENST00000432306.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008927.hg.1 ENST00000432306.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008927.hg.1 ENST00000432306.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008927.hg.1 ENST00000432306.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008927.hg.1 ENST00000432306.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008927.hg.1 ENST00000432306.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008927.hg.1 ENST00000432306.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008927.hg.1 ENST00000432306.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008927.hg.1 ENST00000432306.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008927.hg.1 ENST00000432306.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008927.hg.1 ENST00000472892.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008927.hg.1 ENST00000472892.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008927.hg.1 ENST00000472892.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008927.hg.1 ENST00000472892.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008927.hg.1 ENST00000480057.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008927.hg.1 ENST00000480057.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008927.hg.1 ENST00000480057.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008927.hg.1 ENST00000480057.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008927.hg.1 ENST00000480057.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008927.hg.1 ENST00000483122.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008927.hg.1 ENST00000483122.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008927.hg.1 ENST00000483122.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008927.hg.1 ENST00000483122.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008927.hg.1 ENST00000483122.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008927.hg.1 ENST00000483122.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008927.hg.1 ENST00000489142.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008927.hg.1 ENST00000489142.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008927.hg.1 ENST00000489142.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008927.hg.1 ENST00000489142.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008927.hg.1 ENST00000489142.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008927.hg.1 ENST00000489357.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008927.hg.1 ENST00000489357.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008927.hg.1 ENST00000489357.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008927.hg.1 ENST00000489357.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008927.hg.1 ENST00000489357.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008927.hg.1 ENST00000494728.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008927.hg.1 ENST00000494728.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008927.hg.1 ENST00000494728.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008927.hg.1 ENST00000494728.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008927.hg.1 ENST00000494728.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008927.hg.1 ENST00000496445.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008927.hg.1 ENST00000496445.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008927.hg.1 ENST00000496445.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008927.hg.1 ENST00000496445.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008927.hg.1 ENST00000496445.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008927.hg.1 ENST00000496445.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008927.hg.1 ENST00000496445.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008927.hg.1 ENST00000496445.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008927.hg.1 NM_145206.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000008927.hg.1 NM_145206.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000008927.hg.1 NM_145206.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000008927.hg.1 NM_145206.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000008927.hg.1 NM_145206.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000008927.hg.1 NM_145206.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000008927.hg.1 NM_145206.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000008927.hg.1 NM_145206.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000008927.hg.1 NM_145206.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000008927.hg.1 NM_145206.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000008927.hg.1 NM_145206.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000008927.hg.1 NM_145206.3 Refseq
Affymetrix Human Clariom D Assay TC1000008927.hg.1 ENST00000393077.2 Ensembl
Affymetrix Human Clariom D Assay TC1000008927.hg.1 ENST00000393077.2 Ensembl
Affymetrix Human Clariom D Assay TC1000008927.hg.1 ENST00000393077.2 Ensembl
Affymetrix Human Clariom D Assay TC1000008927.hg.1 ENST00000393077.2 Ensembl
Affymetrix Human Clariom D Assay TC1000008927.hg.1 ENST00000393077.2 Ensembl
Affymetrix Human Clariom D Assay TC1000008927.hg.1 ENST00000393077.2 Ensembl
Affymetrix Human Clariom D Assay TC1000008927.hg.1 ENST00000393077.2 Ensembl
Affymetrix Human Clariom D Assay TC1000008927.hg.1 ENST00000393077.2 Ensembl
Affymetrix Human Clariom D Assay TC1000008927.hg.1 ENST00000393077.2 Ensembl
Affymetrix Human Clariom D Assay TC1000008927.hg.1 ENST00000393077.2 Ensembl
Affymetrix Human Clariom D Assay TC1000008927.hg.1 ENST00000393077.2 Ensembl
Affymetrix Human Clariom D Assay TC1000008927.hg.1 ENST00000432306.5 Ensembl
Affymetrix Human Clariom D Assay TC1000008927.hg.1 ENST00000432306.5 Ensembl
Affymetrix Human Clariom D Assay TC1000008927.hg.1 ENST00000432306.5 Ensembl
Affymetrix Human Clariom D Assay TC1000008927.hg.1 ENST00000432306.5 Ensembl
Affymetrix Human Clariom D Assay TC1000008927.hg.1 ENST00000432306.5 Ensembl
Affymetrix Human Clariom D Assay TC1000008927.hg.1 ENST00000432306.5 Ensembl
Affymetrix Human Clariom D Assay TC1000008927.hg.1 ENST00000432306.5 Ensembl
Affymetrix Human Clariom D Assay TC1000008927.hg.1 ENST00000432306.5 Ensembl
Affymetrix Human Clariom D Assay TC1000008927.hg.1 ENST00000432306.5 Ensembl
Affymetrix Human Clariom D Assay TC1000008927.hg.1 ENST00000432306.5 Ensembl
Affymetrix Human Clariom D Assay TC1000008927.hg.1 ENST00000472892.1 Ensembl
Affymetrix Human Clariom D Assay TC1000008927.hg.1 ENST00000472892.1 Ensembl
Affymetrix Human Clariom D Assay TC1000008927.hg.1 ENST00000472892.1 Ensembl
Affymetrix Human Clariom D Assay TC1000008927.hg.1 ENST00000472892.1 Ensembl
Affymetrix Human Clariom D Assay TC1000008927.hg.1 ENST00000480057.1 Ensembl
Affymetrix Human Clariom D Assay TC1000008927.hg.1 ENST00000480057.1 Ensembl
Affymetrix Human Clariom D Assay TC1000008927.hg.1 ENST00000480057.1 Ensembl
Affymetrix Human Clariom D Assay TC1000008927.hg.1 ENST00000480057.1 Ensembl
Affymetrix Human Clariom D Assay TC1000008927.hg.1 ENST00000480057.1 Ensembl
Affymetrix Human Clariom D Assay TC1000008927.hg.1 ENST00000483122.1 Ensembl
Affymetrix Human Clariom D Assay TC1000008927.hg.1 ENST00000483122.1 Ensembl
Affymetrix Human Clariom D Assay TC1000008927.hg.1 ENST00000483122.1 Ensembl
Affymetrix Human Clariom D Assay TC1000008927.hg.1 ENST00000483122.1 Ensembl
Affymetrix Human Clariom D Assay TC1000008927.hg.1 ENST00000483122.1 Ensembl
Affymetrix Human Clariom D Assay TC1000008927.hg.1 ENST00000483122.1 Ensembl
Affymetrix Human Clariom D Assay TC1000008927.hg.1 ENST00000489142.5 Ensembl
Affymetrix Human Clariom D Assay TC1000008927.hg.1 ENST00000489142.5 Ensembl
Affymetrix Human Clariom D Assay TC1000008927.hg.1 ENST00000489142.5 Ensembl
Affymetrix Human Clariom D Assay TC1000008927.hg.1 ENST00000489142.5 Ensembl
Affymetrix Human Clariom D Assay TC1000008927.hg.1 ENST00000489142.5 Ensembl
Affymetrix Human Clariom D Assay TC1000008927.hg.1 ENST00000489357.5 Ensembl
Affymetrix Human Clariom D Assay TC1000008927.hg.1 ENST00000489357.5 Ensembl
Affymetrix Human Clariom D Assay TC1000008927.hg.1 ENST00000489357.5 Ensembl
Affymetrix Human Clariom D Assay TC1000008927.hg.1 ENST00000489357.5 Ensembl
Affymetrix Human Clariom D Assay TC1000008927.hg.1 ENST00000489357.5 Ensembl
Affymetrix Human Clariom D Assay TC1000008927.hg.1 ENST00000494728.5 Ensembl
Affymetrix Human Clariom D Assay TC1000008927.hg.1 ENST00000494728.5 Ensembl
Affymetrix Human Clariom D Assay TC1000008927.hg.1 ENST00000494728.5 Ensembl
Affymetrix Human Clariom D Assay TC1000008927.hg.1 ENST00000494728.5 Ensembl
Affymetrix Human Clariom D Assay TC1000008927.hg.1 ENST00000494728.5 Ensembl
Affymetrix Human Clariom D Assay TC1000008927.hg.1 ENST00000496445.5 Ensembl
Affymetrix Human Clariom D Assay TC1000008927.hg.1 ENST00000496445.5 Ensembl
Affymetrix Human Clariom D Assay TC1000008927.hg.1 ENST00000496445.5 Ensembl
Affymetrix Human Clariom D Assay TC1000008927.hg.1 ENST00000496445.5 Ensembl
Affymetrix Human Clariom D Assay TC1000008927.hg.1 ENST00000496445.5 Ensembl
Affymetrix Human Clariom D Assay TC1000008927.hg.1 ENST00000496445.5 Ensembl
Affymetrix Human Clariom D Assay TC1000008927.hg.1 ENST00000496445.5 Ensembl
Affymetrix Human Clariom D Assay TC1000008927.hg.1 ENST00000496445.5 Ensembl
Affymetrix Human Clariom D Assay TC1000008927.hg.1 NM_145206.3 Refseq
Affymetrix Human Clariom D Assay TC1000008927.hg.1 NM_145206.3 Refseq
Affymetrix Human Clariom D Assay TC1000008927.hg.1 NM_145206.3 Refseq
Affymetrix Human Clariom D Assay TC1000008927.hg.1 NM_145206.3 Refseq
Affymetrix Human Clariom D Assay TC1000008927.hg.1 NM_145206.3 Refseq
Affymetrix Human Clariom D Assay TC1000008927.hg.1 NM_145206.3 Refseq
Affymetrix Human Clariom D Assay TC1000008927.hg.1 NM_145206.3 Refseq
Affymetrix Human Clariom D Assay TC1000008927.hg.1 NM_145206.3 Refseq
Affymetrix Human Clariom D Assay TC1000008927.hg.1 NM_145206.3 Refseq
Affymetrix Human Clariom D Assay TC1000008927.hg.1 NM_145206.3 Refseq
Affymetrix Human Clariom D Assay TC1000008927.hg.1 NM_145206.3 Refseq
Affymetrix Human Clariom D Assay TC1000008927.hg.1 NM_145206.3 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-7dtsd:80/100.66.76.204:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0