This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs05050163_s1
See other SOCS4 GE Assays ›
Gene Symbol:
SOCS4
Gene Name:
suppressor of cytokine signaling 4
Gene Aliases:
SOCS7
Chromosome Location:
Chr.14: 55027126 - 55049489 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs05050163_s1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: SOCS4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_080867.2 NP_543143.1 1 278 95
NM_199421.1 NP_955453.1 1 278 95
XM_011536425.1 XP_011534727.1 1 273 95
XM_011536426.1 XP_011534728.1 1 273 95
GenBank mRNA AK124083.1 - 1 131 95
BC060790.1 - 1 156 30338646 95
BX161418.1 - 1 174 95
CD109883.1 - 1 305 95
CR933659.1 - 1 168 95

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AK124083.1, BC060790.1, BX161418.1, CR933659.1, NM_080867.2, NM_199421.1, XM_011536425.1, XM_011536426.1 transcripts.

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene SOCS4

Panther Classification:

Molecular Function -

kinase modulator

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of epidermal growth factor-activated receptor activity
protein ubiquitination
cytokine-mediated signaling pathway
positive regulation of proteasomal ubiquitin-dependent protein catabolic process
intracellular signal transduction
regulation of growth
negative regulation of JAK-STAT cascade
protein kinase inhibitor activity
protein binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1400007221.hg.1 ENST00000339298.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400007221.hg.1 ENST00000339298.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400007221.hg.1 ENST00000339298.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400007221.hg.1 ENST00000339298.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400007221.hg.1 ENST00000339298.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400007221.hg.1 ENST00000395472.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400007221.hg.1 ENST00000395472.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400007221.hg.1 ENST00000395472.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400007221.hg.1 ENST00000395472.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400007221.hg.1 ENST00000395472.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400007221.hg.1 ENST00000395472.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400007221.hg.1 ENST00000553735.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400007221.hg.1 ENST00000553735.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400007221.hg.1 ENST00000555846.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400007221.hg.1 ENST00000555846.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400007221.hg.1 ENST00000555846.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400007221.hg.1 ENST00000555846.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400007221.hg.1 ENST00000555846.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400007221.hg.1 ENST00000555846.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400007221.hg.1 ENST00000555846.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400007221.hg.1 NM_080867.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400007221.hg.1 NM_080867.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400007221.hg.1 NM_080867.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400007221.hg.1 NM_080867.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400007221.hg.1 NM_080867.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400007221.hg.1 NM_080867.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400007221.hg.1 NM_080867.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400007221.hg.1 NM_199421.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400007221.hg.1 NM_199421.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400007221.hg.1 NM_199421.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400007221.hg.1 NM_199421.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400007221.hg.1 NM_199421.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400007221.hg.1 NM_199421.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400007221.hg.1 NM_199421.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400007221.hg.1 NM_199421.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400007221.hg.1 NM_199421.1 Refseq
Affymetrix Human Clariom D Assay TC1400007221.hg.1 ENST00000339298.2 Ensembl
Affymetrix Human Clariom D Assay TC1400007221.hg.1 ENST00000339298.2 Ensembl
Affymetrix Human Clariom D Assay TC1400007221.hg.1 ENST00000339298.2 Ensembl
Affymetrix Human Clariom D Assay TC1400007221.hg.1 ENST00000339298.2 Ensembl
Affymetrix Human Clariom D Assay TC1400007221.hg.1 ENST00000339298.2 Ensembl
Affymetrix Human Clariom D Assay TC1400007221.hg.1 ENST00000395472.2 Ensembl
Affymetrix Human Clariom D Assay TC1400007221.hg.1 ENST00000395472.2 Ensembl
Affymetrix Human Clariom D Assay TC1400007221.hg.1 ENST00000395472.2 Ensembl
Affymetrix Human Clariom D Assay TC1400007221.hg.1 ENST00000395472.2 Ensembl
Affymetrix Human Clariom D Assay TC1400007221.hg.1 ENST00000395472.2 Ensembl
Affymetrix Human Clariom D Assay TC1400007221.hg.1 ENST00000395472.2 Ensembl
Affymetrix Human Clariom D Assay TC1400007221.hg.1 ENST00000553735.1 Ensembl
Affymetrix Human Clariom D Assay TC1400007221.hg.1 ENST00000553735.1 Ensembl
Affymetrix Human Clariom D Assay TC1400007221.hg.1 ENST00000555846.1 Ensembl
Affymetrix Human Clariom D Assay TC1400007221.hg.1 ENST00000555846.1 Ensembl
Affymetrix Human Clariom D Assay TC1400007221.hg.1 ENST00000555846.1 Ensembl
Affymetrix Human Clariom D Assay TC1400007221.hg.1 ENST00000555846.1 Ensembl
Affymetrix Human Clariom D Assay TC1400007221.hg.1 ENST00000555846.1 Ensembl
Affymetrix Human Clariom D Assay TC1400007221.hg.1 ENST00000555846.1 Ensembl
Affymetrix Human Clariom D Assay TC1400007221.hg.1 ENST00000555846.1 Ensembl
Affymetrix Human Clariom D Assay TC1400007221.hg.1 NM_080867.2 Refseq
Affymetrix Human Clariom D Assay TC1400007221.hg.1 NM_080867.2 Refseq
Affymetrix Human Clariom D Assay TC1400007221.hg.1 NM_080867.2 Refseq
Affymetrix Human Clariom D Assay TC1400007221.hg.1 NM_080867.2 Refseq
Affymetrix Human Clariom D Assay TC1400007221.hg.1 NM_080867.2 Refseq
Affymetrix Human Clariom D Assay TC1400007221.hg.1 NM_080867.2 Refseq
Affymetrix Human Clariom D Assay TC1400007221.hg.1 NM_080867.2 Refseq
Affymetrix Human Clariom D Assay TC1400007221.hg.1 NM_199421.1 Refseq
Affymetrix Human Clariom D Assay TC1400007221.hg.1 NM_199421.1 Refseq
Affymetrix Human Clariom D Assay TC1400007221.hg.1 NM_199421.1 Refseq
Affymetrix Human Clariom D Assay TC1400007221.hg.1 NM_199421.1 Refseq
Affymetrix Human Clariom D Assay TC1400007221.hg.1 NM_199421.1 Refseq
Affymetrix Human Clariom D Assay TC1400007221.hg.1 NM_199421.1 Refseq
Affymetrix Human Clariom D Assay TC1400007221.hg.1 NM_199421.1 Refseq
Affymetrix Human Clariom D Assay TC1400007221.hg.1 NM_199421.1 Refseq
Affymetrix Human Clariom D Assay TC1400007221.hg.1 NM_199421.1 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-6b469b8bb7-lcddp:80/100.66.76.150:80.
git-commit: a334af76dff23450325448aedefe62379591458a
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.48.1-2026.04.16-1.0