This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs05050874_s1
See other MESP2 GE Assays ›
Gene Symbol:
MESP2
Gene Name:
mesoderm posterior bHLH transcription factor 2
Gene Aliases:
SCDO2, bHLHc6
Chromosome Location:
Chr.15: 89776358 - 89778754 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs05050874_s1
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: MESP2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001039958.1 NP_001035047.1 2 1112 66
GenBank mRNA AL360139.1 - 3 357 66
BC111413.1 - 2 1100 40004357 66

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription, DNA-templated
Notch signaling pathway
multicellular organism development
DNA binding
protein dimerization activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1500008276.hg.1 ENST00000341735.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500008276.hg.1 ENST00000341735.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500008276.hg.1 ENST00000341735.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500008276.hg.1 ENST00000341735.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500008276.hg.1 ENST00000558723.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500008276.hg.1 ENST00000558723.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500008276.hg.1 ENST00000558723.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500008276.hg.1 ENST00000558723.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500008276.hg.1 ENST00000560219.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500008276.hg.1 ENST00000560219.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500008276.hg.1 ENST00000560219.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500008276.hg.1 ENST00000560219.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500008276.hg.1 ENST00000560219.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500008276.hg.1 ENST00000560219.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500008276.hg.1 NM_001039958.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500008276.hg.1 NM_001039958.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500008276.hg.1 NM_001039958.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500008276.hg.1 NM_001039958.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500008276.hg.1 NM_001039958.1 Refseq
Affymetrix Human Clariom D Assay TC1500008276.hg.1 ENST00000341735.3 Ensembl
Affymetrix Human Clariom D Assay TC1500008276.hg.1 ENST00000341735.3 Ensembl
Affymetrix Human Clariom D Assay TC1500008276.hg.1 ENST00000341735.3 Ensembl
Affymetrix Human Clariom D Assay TC1500008276.hg.1 ENST00000341735.3 Ensembl
Affymetrix Human Clariom D Assay TC1500008276.hg.1 ENST00000558723.1 Ensembl
Affymetrix Human Clariom D Assay TC1500008276.hg.1 ENST00000558723.1 Ensembl
Affymetrix Human Clariom D Assay TC1500008276.hg.1 ENST00000558723.1 Ensembl
Affymetrix Human Clariom D Assay TC1500008276.hg.1 ENST00000558723.1 Ensembl
Affymetrix Human Clariom D Assay TC1500008276.hg.1 ENST00000560219.2 Ensembl
Affymetrix Human Clariom D Assay TC1500008276.hg.1 ENST00000560219.2 Ensembl
Affymetrix Human Clariom D Assay TC1500008276.hg.1 ENST00000560219.2 Ensembl
Affymetrix Human Clariom D Assay TC1500008276.hg.1 ENST00000560219.2 Ensembl
Affymetrix Human Clariom D Assay TC1500008276.hg.1 ENST00000560219.2 Ensembl
Affymetrix Human Clariom D Assay TC1500008276.hg.1 ENST00000560219.2 Ensembl
Affymetrix Human Clariom D Assay TC1500008276.hg.1 NM_001039958.1 Refseq
Affymetrix Human Clariom D Assay TC1500008276.hg.1 NM_001039958.1 Refseq
Affymetrix Human Clariom D Assay TC1500008276.hg.1 NM_001039958.1 Refseq
Affymetrix Human Clariom D Assay TC1500008276.hg.1 NM_001039958.1 Refseq
Affymetrix Human Clariom D Assay TC1500008276.hg.1 NM_001039958.1 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-7dtsd:80/100.66.76.204:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0