This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs05050952_s1
See other IGDCC3 GE Assays ›
Gene Symbol:
IGDCC3
Gene Name:
immunoglobulin superfamily DCC subclass member 3
Gene Aliases:
HsT18880, PUNC
Chromosome Location:
Chr.15: 65327127 - 65378040 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs05050952_s1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: IGDCC3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_004884.3 NP_004875.2 14 3271 114
XM_011522241.1 XP_011520543.1 14 3210 114
XM_011522243.1 XP_011520545.1 13 2797 114
XM_011522244.1 XP_011520546.1 13 2786 114
GenBank mRNA AK095529.1 - 14 2666 115

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene IGDCC3

Gene Ontology Categories:

Function(s)

neuromuscular process controlling balance

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0