This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs05050983_s1
See other MESP1 GE Assays ›
Gene Symbol:
MESP1
Gene Name:
mesoderm posterior bHLH transcription factor 1
Gene Aliases:
bHLHc5
Chromosome Location:
Chr.15: 89734772 - 89751309 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs05050983_s1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: MESP1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_018670.3 NP_061140.1 2 909 89
GenBank mRNA AL357533.1 - 2 462 89
AL357534.1 - 2 440 89
AL357535.1 - 1 82 89
AL357570.1 - 2 356 89
AL357571.1 - 2 729 89
BC006219.1 - 2 917 3944350 89
BP212916.1 - 2 479 89

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene MESP1

Gene Ontology Categories:

Function(s) Process(es)

heart looping
secondary heart field specification
embryonic heart tube morphogenesis
cardiac atrium formation
cardiac ventricle formation
sinus venosus morphogenesis
growth involved in heart morphogenesis
cardioblast anterior-lateral migration
transcription from RNA polymerase II promoter
Notch signaling pathway
gastrulation
mesodermal cell migration
neurogenesis
signal transduction involved in regulation of gene expression
positive regulation of Notch signaling pathway involved in heart induction
negative regulation of mesodermal cell fate specification
negative regulation of endodermal cell fate specification
endothelial cell differentiation
positive regulation of Notch signaling pathway
negative regulation of transcription, DNA-templated
positive regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
lateral mesoderm development
positive regulation of striated muscle cell differentiation
cardiac muscle cell differentiation
cardiac cell fate determination
sinoatrial node cell differentiation
cardiac vascular smooth muscle cell differentiation
cardioblast migration to the midline involved in heart field formation
positive regulation of hepatocyte differentiation
positive regulation of heart induction by negative regulation of canonical Wnt signaling pathway
RNA polymerase II core promoter proximal region sequence-specific DNA binding
transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
transcription factor activity, sequence-specific DNA binding
enhancer binding
transcription regulatory region DNA binding
protein dimerization activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1500010404.hg.1 ENST00000300057.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010404.hg.1 ENST00000300057.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010404.hg.1 ENST00000300057.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010404.hg.1 ENST00000300057.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010404.hg.1 ENST00000300057.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010404.hg.1 ENST00000300057.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010404.hg.1 ENST00000300057.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010404.hg.1 ENST00000300057.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010404.hg.1 ENST00000559894.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010404.hg.1 ENST00000559894.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010404.hg.1 ENST00000559894.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010404.hg.1 ENST00000559894.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500010404.hg.1 NM_018670.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500010404.hg.1 NM_018670.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500010404.hg.1 NM_018670.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500010404.hg.1 NM_018670.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500010404.hg.1 NM_018670.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500010404.hg.1 NM_018670.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500010404.hg.1 NM_018670.3 Refseq
Affymetrix Human Clariom D Assay TC1500010404.hg.1 ENST00000300057.4 Ensembl
Affymetrix Human Clariom D Assay TC1500010404.hg.1 ENST00000300057.4 Ensembl
Affymetrix Human Clariom D Assay TC1500010404.hg.1 ENST00000300057.4 Ensembl
Affymetrix Human Clariom D Assay TC1500010404.hg.1 ENST00000300057.4 Ensembl
Affymetrix Human Clariom D Assay TC1500010404.hg.1 ENST00000300057.4 Ensembl
Affymetrix Human Clariom D Assay TC1500010404.hg.1 ENST00000300057.4 Ensembl
Affymetrix Human Clariom D Assay TC1500010404.hg.1 ENST00000300057.4 Ensembl
Affymetrix Human Clariom D Assay TC1500010404.hg.1 ENST00000300057.4 Ensembl
Affymetrix Human Clariom D Assay TC1500010404.hg.1 ENST00000559894.1 Ensembl
Affymetrix Human Clariom D Assay TC1500010404.hg.1 ENST00000559894.1 Ensembl
Affymetrix Human Clariom D Assay TC1500010404.hg.1 ENST00000559894.1 Ensembl
Affymetrix Human Clariom D Assay TC1500010404.hg.1 ENST00000559894.1 Ensembl
Affymetrix Human Clariom D Assay TC1500010404.hg.1 NM_018670.3 Refseq
Affymetrix Human Clariom D Assay TC1500010404.hg.1 NM_018670.3 Refseq
Affymetrix Human Clariom D Assay TC1500010404.hg.1 NM_018670.3 Refseq
Affymetrix Human Clariom D Assay TC1500010404.hg.1 NM_018670.3 Refseq
Affymetrix Human Clariom D Assay TC1500010404.hg.1 NM_018670.3 Refseq
Affymetrix Human Clariom D Assay TC1500010404.hg.1 NM_018670.3 Refseq
Affymetrix Human Clariom D Assay TC1500010404.hg.1 NM_018670.3 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-2k58s:80/100.66.76.192:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0