This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs05050997_s1
See other FURIN GE Assays ›
Gene Symbol:
FURIN
Gene Name:
furin, paired basic amino acid cleaving enzyme
Gene Aliases:
FUR, PACE, PCSK3, SPC1
Chromosome Location:
Chr.15: 90868592 - 90883458 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs05050997_s1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: FURIN
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001289823.1 NP_001276752.1 16 3257 74
NM_001289824.1 NP_001276753.1 16 3411 74
NM_002569.3 NP_002560.1 16 3310 74
GenBank mRNA BC008295.1 - 1 498 74
BC012181.1 - 15 3080 4550620 74
X17094.1 - 16 3247 74

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene FURIN

Panther Classification:

Molecular Function -

serine protease

Gene Ontology Categories:

Function(s) Process(es)

signal peptide processing
transforming growth factor beta receptor signaling pathway
aging
cell proliferation
negative regulation of endopeptidase activity
protein processing
peptide hormone processing
viral life cycle
viral protein processing
extracellular matrix disassembly
extracellular matrix organization
positive regulation of cell migration
positive regulation of transforming growth factor beta receptor signaling pathway
collagen catabolic process
nerve growth factor processing
negative regulation of low-density lipoprotein particle receptor catabolic process
nerve growth factor production
negative regulation of nerve growth factor production
negative regulation of transforming growth factor beta1 production
secretion by cell
regulation of protein catabolic process
peptide biosynthetic process
cellular protein metabolic process
regulation of low-density lipoprotein particle receptor biosynthetic process
positive regulation of membrane protein ectodomain proteolysis
regulation of endopeptidase activity
protease binding
endopeptidase activity
serine-type endopeptidase activity
serine-type endopeptidase inhibitor activity
protein binding
peptidase activity
peptide binding
metal ion binding
nerve growth factor binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-8cb8z:80/100.66.72.77:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0