| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009172.hg.1 |
ENST00000402693.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009172.hg.1 |
ENST00000402693.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009172.hg.1 |
ENST00000402693.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009172.hg.1 |
ENST00000402693.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009172.hg.1 |
ENST00000402693.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009172.hg.1 |
ENST00000402693.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009172.hg.1 |
ENST00000402693.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009172.hg.1 |
ENST00000402693.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009172.hg.1 |
ENST00000402693.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009172.hg.1 |
ENST00000402693.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009172.hg.1 |
ENST00000605173.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009172.hg.1 |
ENST00000605173.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009172.hg.1 |
ENST00000605173.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009172.hg.1 |
ENST00000605173.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009172.hg.1 |
ENST00000605173.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009172.hg.1 |
NM_001174118.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009172.hg.1 |
NM_001174118.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009172.hg.1 |
NM_001174118.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009172.hg.1 |
NM_001174118.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009172.hg.1 |
NM_001174118.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009172.hg.1 |
NM_001174118.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009172.hg.1 |
NM_001174118.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009172.hg.1 |
NM_001174118.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009172.hg.1 |
NM_001174118.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009172.hg.1 |
NM_203304.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009172.hg.1 |
NM_203304.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009172.hg.1 |
NM_203304.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009172.hg.1 |
NM_203304.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009172.hg.1 |
NM_203304.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009172.hg.1 |
NM_203304.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009172.hg.1 |
NM_203304.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009172.hg.1 |
NM_203304.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009172.hg.1 |
NM_203304.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009172.hg.1 |
NM_203304.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900009172.hg.1 |
ENST00000402693.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009172.hg.1 |
ENST00000402693.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009172.hg.1 |
ENST00000402693.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009172.hg.1 |
ENST00000402693.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009172.hg.1 |
ENST00000402693.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009172.hg.1 |
ENST00000402693.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009172.hg.1 |
ENST00000402693.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009172.hg.1 |
ENST00000402693.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009172.hg.1 |
ENST00000402693.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009172.hg.1 |
ENST00000402693.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009172.hg.1 |
ENST00000605173.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009172.hg.1 |
ENST00000605173.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009172.hg.1 |
ENST00000605173.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009172.hg.1 |
ENST00000605173.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009172.hg.1 |
ENST00000605173.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009172.hg.1 |
NM_001174118.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900009172.hg.1 |
NM_001174118.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900009172.hg.1 |
NM_001174118.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900009172.hg.1 |
NM_001174118.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900009172.hg.1 |
NM_001174118.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900009172.hg.1 |
NM_001174118.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900009172.hg.1 |
NM_001174118.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900009172.hg.1 |
NM_001174118.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900009172.hg.1 |
NM_001174118.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900009172.hg.1 |
NM_203304.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900009172.hg.1 |
NM_203304.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900009172.hg.1 |
NM_203304.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900009172.hg.1 |
NM_203304.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900009172.hg.1 |
NM_203304.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900009172.hg.1 |
NM_203304.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900009172.hg.1 |
NM_203304.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900009172.hg.1 |
NM_203304.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900009172.hg.1 |
NM_203304.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900009172.hg.1 |
NM_203304.3 |
Refseq |