This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs05053828_s1
See other RAMP2 GE Assays ›
Gene Symbol:
RAMP2
Gene Name:
receptor activity modifying protein 2
Gene Aliases:
-
Chromosome Location:
Chr.17: 42761194 - 42763041 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs05053828_s1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: RAMP2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_005854.2 NP_005845.2 4 695 63
GenBank mRNA AJ001015.1 - 4 694 63
AL548945.3 - 4 701 63
BC027975.1 - 4 644 5229167 63

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene RAMP2

Panther Classification:

Molecular Function -

transmembrane signal receptor

Gene Ontology Categories:

Function(s) Process(es)

angiogenesis
vasculogenesis
response to hypoxia
sprouting angiogenesis
cAMP biosynthetic process
calcium ion transport
intracellular protein transport
G-protein coupled receptor signaling pathway
heart development
female pregnancy
regulation of blood pressure
regulation of G-protein coupled receptor protein signaling pathway
positive regulation of gene expression
protein transport
positive regulation of cAMP biosynthetic process
receptor internalization
response to estradiol
response to progesterone
cellular response to hormone stimulus
adherens junction assembly
cellular response to vascular endothelial growth factor stimulus
negative regulation of vascular permeability
positive regulation of angiogenesis
bicellular tight junction assembly
basement membrane assembly
protein localization to plasma membrane
vascular smooth muscle cell development
negative regulation of endothelial cell apoptotic process
positive regulation of vasculogenesis
protein binding
protein transporter activity
coreceptor activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1700007913.hg.1 ENST00000253796.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007913.hg.1 ENST00000253796.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007913.hg.1 ENST00000253796.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007913.hg.1 ENST00000253796.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007913.hg.1 ENST00000253796.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007913.hg.1 ENST00000253796.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007913.hg.1 ENST00000253796.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007913.hg.1 ENST00000253796.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007913.hg.1 ENST00000587142.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007913.hg.1 ENST00000587142.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007913.hg.1 ENST00000587142.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007913.hg.1 ENST00000587142.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007913.hg.1 ENST00000587142.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007913.hg.1 ENST00000588576.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007913.hg.1 ENST00000588576.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007913.hg.1 ENST00000588576.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007913.hg.1 ENST00000588576.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007913.hg.1 ENST00000588576.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007913.hg.1 ENST00000588928.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007913.hg.1 ENST00000588928.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007913.hg.1 ENST00000588928.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007913.hg.1 ENST00000588928.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007913.hg.1 ENST00000588928.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007913.hg.1 ENST00000589683.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007913.hg.1 ENST00000589683.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007913.hg.1 ENST00000589683.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007913.hg.1 ENST00000589683.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007913.hg.1 ENST00000589683.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007913.hg.1 ENST00000589683.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007913.hg.1 ENST00000589683.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007913.hg.1 ENST00000589683.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007913.hg.1 ENST00000591972.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007913.hg.1 ENST00000591972.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007913.hg.1 NM_005854.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700007913.hg.1 NM_005854.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700007913.hg.1 NM_005854.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700007913.hg.1 NM_005854.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700007913.hg.1 NM_005854.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700007913.hg.1 NM_005854.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700007913.hg.1 NM_005854.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700007913.hg.1 NM_005854.2 Refseq
Affymetrix Human Clariom D Assay TC1700007913.hg.1 ENST00000253796.9 Ensembl
Affymetrix Human Clariom D Assay TC1700007913.hg.1 ENST00000253796.9 Ensembl
Affymetrix Human Clariom D Assay TC1700007913.hg.1 ENST00000253796.9 Ensembl
Affymetrix Human Clariom D Assay TC1700007913.hg.1 ENST00000253796.9 Ensembl
Affymetrix Human Clariom D Assay TC1700007913.hg.1 ENST00000253796.9 Ensembl
Affymetrix Human Clariom D Assay TC1700007913.hg.1 ENST00000253796.9 Ensembl
Affymetrix Human Clariom D Assay TC1700007913.hg.1 ENST00000253796.9 Ensembl
Affymetrix Human Clariom D Assay TC1700007913.hg.1 ENST00000253796.9 Ensembl
Affymetrix Human Clariom D Assay TC1700007913.hg.1 ENST00000587142.5 Ensembl
Affymetrix Human Clariom D Assay TC1700007913.hg.1 ENST00000587142.5 Ensembl
Affymetrix Human Clariom D Assay TC1700007913.hg.1 ENST00000587142.5 Ensembl
Affymetrix Human Clariom D Assay TC1700007913.hg.1 ENST00000587142.5 Ensembl
Affymetrix Human Clariom D Assay TC1700007913.hg.1 ENST00000587142.5 Ensembl
Affymetrix Human Clariom D Assay TC1700007913.hg.1 ENST00000588576.1 Ensembl
Affymetrix Human Clariom D Assay TC1700007913.hg.1 ENST00000588576.1 Ensembl
Affymetrix Human Clariom D Assay TC1700007913.hg.1 ENST00000588576.1 Ensembl
Affymetrix Human Clariom D Assay TC1700007913.hg.1 ENST00000588576.1 Ensembl
Affymetrix Human Clariom D Assay TC1700007913.hg.1 ENST00000588576.1 Ensembl
Affymetrix Human Clariom D Assay TC1700007913.hg.1 ENST00000588928.1 Ensembl
Affymetrix Human Clariom D Assay TC1700007913.hg.1 ENST00000588928.1 Ensembl
Affymetrix Human Clariom D Assay TC1700007913.hg.1 ENST00000588928.1 Ensembl
Affymetrix Human Clariom D Assay TC1700007913.hg.1 ENST00000588928.1 Ensembl
Affymetrix Human Clariom D Assay TC1700007913.hg.1 ENST00000588928.1 Ensembl
Affymetrix Human Clariom D Assay TC1700007913.hg.1 ENST00000589683.5 Ensembl
Affymetrix Human Clariom D Assay TC1700007913.hg.1 ENST00000589683.5 Ensembl
Affymetrix Human Clariom D Assay TC1700007913.hg.1 ENST00000589683.5 Ensembl
Affymetrix Human Clariom D Assay TC1700007913.hg.1 ENST00000589683.5 Ensembl
Affymetrix Human Clariom D Assay TC1700007913.hg.1 ENST00000589683.5 Ensembl
Affymetrix Human Clariom D Assay TC1700007913.hg.1 ENST00000589683.5 Ensembl
Affymetrix Human Clariom D Assay TC1700007913.hg.1 ENST00000589683.5 Ensembl
Affymetrix Human Clariom D Assay TC1700007913.hg.1 ENST00000589683.5 Ensembl
Affymetrix Human Clariom D Assay TC1700007913.hg.1 ENST00000591972.5 Ensembl
Affymetrix Human Clariom D Assay TC1700007913.hg.1 ENST00000591972.5 Ensembl
Affymetrix Human Clariom D Assay TC1700007913.hg.1 NM_005854.2 Refseq
Affymetrix Human Clariom D Assay TC1700007913.hg.1 NM_005854.2 Refseq
Affymetrix Human Clariom D Assay TC1700007913.hg.1 NM_005854.2 Refseq
Affymetrix Human Clariom D Assay TC1700007913.hg.1 NM_005854.2 Refseq
Affymetrix Human Clariom D Assay TC1700007913.hg.1 NM_005854.2 Refseq
Affymetrix Human Clariom D Assay TC1700007913.hg.1 NM_005854.2 Refseq
Affymetrix Human Clariom D Assay TC1700007913.hg.1 NM_005854.2 Refseq
Affymetrix Human Clariom D Assay TC1700007913.hg.1 NM_005854.2 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-hk65g:80/100.66.77.69:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0