This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs05053971_s1
See other GLTPD2 GE Assays ›
Gene Symbol:
GLTPD2 LOC284014
Gene Name:
glycolipid transfer protein domain containing 2
uncharacterized LOC284014
Gene Aliases:
-
-
Chromosome Location:
Chr.17: 4788916 - 4790589 on Build GRCh38
Chromosome Location:
-
Species:
Human
Species Specific ID (Flybase ID):
-
-
Assay ID Hs05053971_s1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: GLTPD2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_005256626.3 XP_005256683.1 1 198 85
Gene Symbol: LOC284014
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK057919.1 - 1 85
AK095567.1 - 1 85

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 3' Most Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Panther Classification:

Molecular Function -

transfer/carrier protein

Gene Ontology Categories:

Function(s) Process(es)

glycolipid transport
glycolipid transporter activity
glycolipid binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1700006643.hg.1 ENST00000331264.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700006643.hg.1 ENST00000331264.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700006643.hg.1 ENST00000331264.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700006643.hg.1 ENST00000331264.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700006643.hg.1 ENST00000331264.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700006643.hg.1 ENST00000331264.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700006643.hg.1 ENST00000623798.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700006643.hg.1 ENST00000623798.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700006643.hg.1 ENST00000623798.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700006643.hg.1 ENST00000623798.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700006643.hg.1 NM_001014985.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700006643.hg.1 NM_001014985.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700006643.hg.1 NM_001014985.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700006643.hg.1 NM_001014985.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700006643.hg.1 NM_001014985.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700006643.hg.1 NM_001014985.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700006643.hg.1 NM_001014985.2 Refseq
Affymetrix Human Clariom D Assay TC1700006643.hg.1 ENST00000331264.7 Ensembl
Affymetrix Human Clariom D Assay TC1700006643.hg.1 ENST00000331264.7 Ensembl
Affymetrix Human Clariom D Assay TC1700006643.hg.1 ENST00000331264.7 Ensembl
Affymetrix Human Clariom D Assay TC1700006643.hg.1 ENST00000331264.7 Ensembl
Affymetrix Human Clariom D Assay TC1700006643.hg.1 ENST00000331264.7 Ensembl
Affymetrix Human Clariom D Assay TC1700006643.hg.1 ENST00000331264.7 Ensembl
Affymetrix Human Clariom D Assay TC1700006643.hg.1 ENST00000623798.1 Ensembl
Affymetrix Human Clariom D Assay TC1700006643.hg.1 ENST00000623798.1 Ensembl
Affymetrix Human Clariom D Assay TC1700006643.hg.1 ENST00000623798.1 Ensembl
Affymetrix Human Clariom D Assay TC1700006643.hg.1 ENST00000623798.1 Ensembl
Affymetrix Human Clariom D Assay TC1700006643.hg.1 NM_001014985.2 Refseq
Affymetrix Human Clariom D Assay TC1700006643.hg.1 NM_001014985.2 Refseq
Affymetrix Human Clariom D Assay TC1700006643.hg.1 NM_001014985.2 Refseq
Affymetrix Human Clariom D Assay TC1700006643.hg.1 NM_001014985.2 Refseq
Affymetrix Human Clariom D Assay TC1700006643.hg.1 NM_001014985.2 Refseq
Affymetrix Human Clariom D Assay TC1700006643.hg.1 NM_001014985.2 Refseq
Affymetrix Human Clariom D Assay TC1700006643.hg.1 NM_001014985.2 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-l4hk6:80/100.66.76.55:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0