This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs05054486_s1
See other SS18 GE Assays ›
Gene Symbol:
SS18
Gene Name:
SS18, nBAF chromatin remodeling complex subunit
Gene Aliases:
SSXT, SYT
Chromosome Location:
Chr.18: 26016253 - 26091217 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs05054486_s1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: SS18
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001007559.2 NP_001007560.1 10 1233 67
NM_001308201.1 NP_001295130.1 10 1208 67
NM_005637.3 NP_005628.2 9 1140 67
XM_006722527.2 XP_006722590.1 11 1259 67
XM_011526145.1 XP_011524447.1 10 1201 67
XM_011526147.2 XP_011524449.1 12 1484 67
XM_011526148.2 XP_011524450.1 12 1471 67
XM_011526149.2 XP_011524451.1 12 1496 67
XM_011526150.2 XP_011524452.1 11 1356 67
XM_011526151.2 XP_011524453.1 12 1485 67
XM_011526152.2 XP_011524454.1 10 1263 67
GenBank mRNA AF244972.1 - 10 1170 67
AF343880.1 - 10 1167 67
AK289510.1 - 10 1211 67
AK296949.1 - 10 1191 67
AK299082.1 - 9 1101 67
AK301504.1 - 9 1145 67
AK304305.1 - 10 1256 67
AK316302.1 - 9 1090 67
AK316338.1 - 10 1209 67
BC096221.1 - 9 1077 40004908 67
BC096222.1 - 10 1170 40004909 67
BC096223.1 - 10 1170 40004911 67
BC096224.1 - 9 1077 40004912 67
CR542088.1 - 9 1074 67
CR542103.1 - 9 1074 67
X79201.1 - 9 1077 67

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene SS18

Panther Classification:

Molecular Function -

transcription cofactor

Gene Ontology Categories:

Function(s) Process(es)

microtubule cytoskeleton organization
cell morphogenesis
transcription, DNA-templated
intracellular signal transduction
response to drug
positive regulation of transcription from RNA polymerase II promoter
ephrin receptor signaling pathway
neuronal stem cell population maintenance
protein binding
ligand-dependent nuclear receptor transcription coactivator activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-gm74s:80/100.66.78.154:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline