This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs05057556_s1
See other STAU1 GE Assays ›
Gene Symbol:
STAU1
Gene Name:
staufen double-stranded RNA binding protein 1
Gene Aliases:
PPP1R150, STAU
Chromosome Location:
Chr.20: 49113339 - 49219295 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs05057556_s1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: STAU1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001037328.2 NP_001032405.1 7 918 83
NM_001319134.1 NP_001306063.1 6 795 83
NM_001319135.1 NP_001306064.1 8 977 83
NM_001322927.1 NP_001309856.1 5 720 83
NM_001322928.1 NP_001309857.1 8 1064 83
NM_001322929.1 NP_001309858.1 8 1189 83
NM_001322930.1 NP_001309859.1 6 991 83
NM_001322931.1 NP_001309860.1 9 1169 83
NM_001322932.1 NP_001309861.1 6 1009 83
NM_001322933.1 NP_001309862.1 7 1084 83
NM_004602.3 NP_004593.2 7 900 83
NM_017452.3 NP_059346.2 6 777 83
NM_017453.3 NP_059347.2 7 1066 83
NM_017454.3 NP_059348.2 5 702 83
XM_005260527.1 XP_005260584.1 7 967 83
XM_006723865.1 XP_006723928.1 8 1204 83
XM_011529015.1 XP_011527317.1 9 1317 83
XM_017028028.1 XP_016883517.1 8 1043 83
GenBank mRNA AB209561.1 - 7 931 83
AF061938.1 - 6 771 83
AF061939.1 - 7 1060 83
AF061940.1 - 5 696 83
AF061941.1 - 7 903 83
AJ132258.1 - 4 444 83
AK025519.1 - 6 740 83
AK057739.1 - 7 1023 83
AK091405.1 - 3 256 83
AK291487.1 - 7 981 83
AK292859.1 - 6 562 83
AL136601.2 - 6 760 83
AY529074.1 - 6 613 83
AY546099.1 - 6 789 83
BC001893.1 - 7 722 83
BC010169.2 - 7 722 83
BC050432.1 - 6 804 6047265 83
BC095397.1 - 6 749 30528051 83

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AK091405.1 transcripts.

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene STAU1

Gene Ontology Categories:

Function(s) Process(es)

cellular response to oxidative stress
positive regulation of viral genome replication
positive regulation by virus of viral protein levels in host cell
positive regulation of long-term synaptic potentiation
double-stranded RNA binding
protein binding
protein phosphatase 1 binding
poly(A) RNA binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-6s775:80/100.66.79.102:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0