This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs05058126_s1
See other BACE2 GE Assays ›
Gene Symbol:
BACE2
Gene Name:
beta-site APP-cleaving enzyme 2
Gene Aliases:
AEPLC, ALP56, ASP1, ASP21, BAE2, CDA13, CEAP1, DRAP
Chromosome Location:
Chr.21: 41167801 - 41282534 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs05058126_s1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: BACE2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_012105.4 NP_036237.2 9 1882 106
NM_138991.2 NP_620476.1 8 1732 106
NM_138992.2 NP_620477.1 8 1713 106
XM_017028314.1 XP_016883803.1 9 1554 106
GenBank mRNA AF050171.1 - 9 1527 106
AF117892.1 - 9 1519 106
AF178532.1 - 9 1882 106
AF188276.1 - 8 1732 106
AF188277.1 - 8 1713 106
AF200192.1 - 9 1496 106
AF200342.1 - 9 1419 106
AF204944.1 - 9 1509 106
AF212252.1 - 9 1240 106
AK075539.1 - 10 1376 106
AK292056.1 - 9 1555 106
AY358927.1 - 9 1512 106
AY429521.1 - 2 222 106
BC014453.1 - 9 1510 4868925 106

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene BACE2

Panther Classification:

Molecular Function -

protease

Gene Ontology Categories:

Function(s) Process(es)

proteolysis
membrane protein ectodomain proteolysis
peptide hormone processing
protein catabolic process
negative regulation of amyloid precursor protein biosynthetic process
beta-amyloid metabolic process
beta-amyloid binding
aspartic-type endopeptidase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000328735.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000328735.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000328735.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000328735.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000328735.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000328735.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000328735.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000328735.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000328735.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000328735.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000328735.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000328735.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000328735.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000328735.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000328735.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000328735.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000330333.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000330333.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000330333.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000330333.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000330333.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000330333.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000330333.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000330333.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000330333.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000330333.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000330333.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000330333.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000330333.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000330333.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000330333.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000330333.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000330333.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000347667.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000347667.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000347667.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000347667.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000347667.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000347667.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000347667.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000347667.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000347667.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000347667.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000347667.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000347667.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000347667.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000347667.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000347667.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000347667.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000347667.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000347667.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000463674.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000463674.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000463674.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000463674.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000463674.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000463674.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000463674.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000463674.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000463674.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000463674.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000465326.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000465326.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000465326.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000465326.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000465326.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000465326.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000465326.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000465326.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000466122.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000466122.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000466122.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000466122.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000466122.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000466122.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000466122.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000466122.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000466122.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000466122.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000466122.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000466122.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000466122.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000470864.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000470864.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000470864.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000470864.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000475618.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000475618.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000487994.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000487994.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000487994.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000487994.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000487994.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000487994.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000487994.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000487994.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000487994.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000491838.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000491838.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000491838.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000491838.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000491838.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000491838.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 ENST00000491838.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 NM_012105.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 NM_012105.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 NM_012105.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 NM_012105.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 NM_012105.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 NM_012105.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 NM_012105.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 NM_012105.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 NM_012105.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 NM_012105.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 NM_012105.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 NM_012105.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 NM_012105.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 NM_012105.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 NM_012105.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 NM_012105.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 NM_012105.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 NM_012105.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 NM_012105.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 NM_138991.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 NM_138991.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 NM_138991.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 NM_138991.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 NM_138991.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 NM_138991.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 NM_138991.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 NM_138991.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 NM_138991.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 NM_138991.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 NM_138991.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 NM_138991.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 NM_138991.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 NM_138991.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 NM_138991.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 NM_138991.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 NM_138991.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 NM_138991.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 NM_138992.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 NM_138992.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 NM_138992.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 NM_138992.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 NM_138992.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 NM_138992.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 NM_138992.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 NM_138992.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 NM_138992.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 NM_138992.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 NM_138992.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 NM_138992.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 NM_138992.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 NM_138992.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 NM_138992.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 NM_138992.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 NM_138992.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100007198.hg.1 NM_138992.2 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-6b469b8bb7-b97mm:80/100.66.72.101:80.
git-commit: a334af76dff23450325448aedefe62379591458a
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.48.1-2026.04.16-1.0