This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs05061493_ft
See other KIAA1549-BRAF FUSION GE Assays ›
Gene Symbol:
KIAA1549-BRAF fusion
Gene Name:
FUSION
Gene Aliases:
-
Chromosome Location:
-
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs05061493_ft
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: KIAA1549-BRAF fusion
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA OM_KIAA1549-BRAF_K13B9.0 - 61 112

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Fusion Homo sapiens

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0700012755.hg.1 ENST00000422774.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012755.hg.1 ENST00000422774.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012755.hg.1 ENST00000422774.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012755.hg.1 ENST00000422774.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012755.hg.1 ENST00000422774.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012755.hg.1 ENST00000422774.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012755.hg.1 ENST00000422774.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012755.hg.1 ENST00000422774.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012755.hg.1 ENST00000422774.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012755.hg.1 ENST00000422774.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012755.hg.1 ENST00000422774.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012755.hg.1 ENST00000422774.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012755.hg.1 ENST00000422774.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012755.hg.1 ENST00000422774.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012755.hg.1 ENST00000422774.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012755.hg.1 ENST00000422774.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012755.hg.1 ENST00000422774.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012755.hg.1 ENST00000422774.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012755.hg.1 ENST00000422774.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012755.hg.1 ENST00000422774.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012755.hg.1 ENST00000422774.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012755.hg.1 ENST00000422774.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012755.hg.1 ENST00000422774.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012755.hg.1 ENST00000440172.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012755.hg.1 ENST00000440172.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012755.hg.1 ENST00000440172.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012755.hg.1 ENST00000440172.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012755.hg.1 ENST00000440172.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012755.hg.1 ENST00000440172.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012755.hg.1 ENST00000440172.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012755.hg.1 ENST00000440172.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012755.hg.1 ENST00000440172.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012755.hg.1 ENST00000440172.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012755.hg.1 ENST00000440172.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012755.hg.1 ENST00000440172.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012755.hg.1 ENST00000440172.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012755.hg.1 ENST00000440172.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012755.hg.1 ENST00000440172.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012755.hg.1 ENST00000440172.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012755.hg.1 ENST00000440172.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012755.hg.1 ENST00000440172.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012755.hg.1 ENST00000440172.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012755.hg.1 ENST00000440172.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012755.hg.1 ENST00000440172.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012755.hg.1 ENST00000440172.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012755.hg.1 ENST00000440172.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012755.hg.1 NM_001164665.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700012755.hg.1 NM_001164665.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700012755.hg.1 NM_001164665.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700012755.hg.1 NM_001164665.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700012755.hg.1 NM_001164665.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700012755.hg.1 NM_001164665.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700012755.hg.1 NM_001164665.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700012755.hg.1 NM_001164665.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700012755.hg.1 NM_001164665.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700012755.hg.1 NM_001164665.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700012755.hg.1 NM_001164665.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700012755.hg.1 NM_001164665.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700012755.hg.1 NM_001164665.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700012755.hg.1 NM_001164665.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700012755.hg.1 NM_001164665.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700012755.hg.1 NM_001164665.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700012755.hg.1 NM_001164665.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700012755.hg.1 NM_001164665.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700012755.hg.1 NM_001164665.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700012755.hg.1 NM_001164665.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700012755.hg.1 NM_001164665.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700012755.hg.1 NM_001164665.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700012755.hg.1 NM_001164665.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700012755.hg.1 NM_001164665.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700012755.hg.1 NM_020910.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700012755.hg.1 NM_020910.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700012755.hg.1 NM_020910.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700012755.hg.1 NM_020910.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700012755.hg.1 NM_020910.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700012755.hg.1 NM_020910.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700012755.hg.1 NM_020910.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700012755.hg.1 NM_020910.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700012755.hg.1 NM_020910.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700012755.hg.1 NM_020910.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700012755.hg.1 NM_020910.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700012755.hg.1 NM_020910.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700012755.hg.1 NM_020910.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700012755.hg.1 NM_020910.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700012755.hg.1 NM_020910.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700012755.hg.1 NM_020910.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700012755.hg.1 NM_020910.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700012755.hg.1 NM_020910.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700012755.hg.1 NM_020910.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700012755.hg.1 NM_020910.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700012755.hg.1 NM_020910.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700012755.hg.1 NM_020910.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700012755.hg.1 NM_020910.2 Refseq
Affymetrix Human Clariom D Assay TC0700012755.hg.1 ENST00000422774.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012755.hg.1 ENST00000422774.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012755.hg.1 ENST00000422774.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012755.hg.1 ENST00000422774.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012755.hg.1 ENST00000422774.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012755.hg.1 ENST00000422774.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012755.hg.1 ENST00000422774.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012755.hg.1 ENST00000422774.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012755.hg.1 ENST00000422774.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012755.hg.1 ENST00000422774.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012755.hg.1 ENST00000422774.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012755.hg.1 ENST00000422774.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012755.hg.1 ENST00000422774.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012755.hg.1 ENST00000422774.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012755.hg.1 ENST00000422774.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012755.hg.1 ENST00000422774.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012755.hg.1 ENST00000422774.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012755.hg.1 ENST00000422774.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012755.hg.1 ENST00000422774.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012755.hg.1 ENST00000422774.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012755.hg.1 ENST00000422774.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012755.hg.1 ENST00000422774.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012755.hg.1 ENST00000422774.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012755.hg.1 ENST00000440172.5 Ensembl
Affymetrix Human Clariom D Assay TC0700012755.hg.1 ENST00000440172.5 Ensembl
Affymetrix Human Clariom D Assay TC0700012755.hg.1 ENST00000440172.5 Ensembl
Affymetrix Human Clariom D Assay TC0700012755.hg.1 ENST00000440172.5 Ensembl
Affymetrix Human Clariom D Assay TC0700012755.hg.1 ENST00000440172.5 Ensembl
Affymetrix Human Clariom D Assay TC0700012755.hg.1 ENST00000440172.5 Ensembl
Affymetrix Human Clariom D Assay TC0700012755.hg.1 ENST00000440172.5 Ensembl
Affymetrix Human Clariom D Assay TC0700012755.hg.1 ENST00000440172.5 Ensembl
Affymetrix Human Clariom D Assay TC0700012755.hg.1 ENST00000440172.5 Ensembl
Affymetrix Human Clariom D Assay TC0700012755.hg.1 ENST00000440172.5 Ensembl
Affymetrix Human Clariom D Assay TC0700012755.hg.1 ENST00000440172.5 Ensembl
Affymetrix Human Clariom D Assay TC0700012755.hg.1 ENST00000440172.5 Ensembl
Affymetrix Human Clariom D Assay TC0700012755.hg.1 ENST00000440172.5 Ensembl
Affymetrix Human Clariom D Assay TC0700012755.hg.1 ENST00000440172.5 Ensembl
Affymetrix Human Clariom D Assay TC0700012755.hg.1 ENST00000440172.5 Ensembl
Affymetrix Human Clariom D Assay TC0700012755.hg.1 ENST00000440172.5 Ensembl
Affymetrix Human Clariom D Assay TC0700012755.hg.1 ENST00000440172.5 Ensembl
Affymetrix Human Clariom D Assay TC0700012755.hg.1 ENST00000440172.5 Ensembl
Affymetrix Human Clariom D Assay TC0700012755.hg.1 ENST00000440172.5 Ensembl
Affymetrix Human Clariom D Assay TC0700012755.hg.1 ENST00000440172.5 Ensembl
Affymetrix Human Clariom D Assay TC0700012755.hg.1 ENST00000440172.5 Ensembl
Affymetrix Human Clariom D Assay TC0700012755.hg.1 ENST00000440172.5 Ensembl
Affymetrix Human Clariom D Assay TC0700012755.hg.1 ENST00000440172.5 Ensembl
Affymetrix Human Clariom D Assay TC0700012755.hg.1 NM_001164665.1 Refseq
Affymetrix Human Clariom D Assay TC0700012755.hg.1 NM_001164665.1 Refseq
Affymetrix Human Clariom D Assay TC0700012755.hg.1 NM_001164665.1 Refseq
Affymetrix Human Clariom D Assay TC0700012755.hg.1 NM_001164665.1 Refseq
Affymetrix Human Clariom D Assay TC0700012755.hg.1 NM_001164665.1 Refseq
Affymetrix Human Clariom D Assay TC0700012755.hg.1 NM_001164665.1 Refseq
Affymetrix Human Clariom D Assay TC0700012755.hg.1 NM_001164665.1 Refseq
Affymetrix Human Clariom D Assay TC0700012755.hg.1 NM_001164665.1 Refseq
Affymetrix Human Clariom D Assay TC0700012755.hg.1 NM_001164665.1 Refseq
Affymetrix Human Clariom D Assay TC0700012755.hg.1 NM_001164665.1 Refseq
Affymetrix Human Clariom D Assay TC0700012755.hg.1 NM_001164665.1 Refseq
Affymetrix Human Clariom D Assay TC0700012755.hg.1 NM_001164665.1 Refseq
Affymetrix Human Clariom D Assay TC0700012755.hg.1 NM_001164665.1 Refseq
Affymetrix Human Clariom D Assay TC0700012755.hg.1 NM_001164665.1 Refseq
Affymetrix Human Clariom D Assay TC0700012755.hg.1 NM_001164665.1 Refseq
Affymetrix Human Clariom D Assay TC0700012755.hg.1 NM_001164665.1 Refseq
Affymetrix Human Clariom D Assay TC0700012755.hg.1 NM_001164665.1 Refseq
Affymetrix Human Clariom D Assay TC0700012755.hg.1 NM_001164665.1 Refseq
Affymetrix Human Clariom D Assay TC0700012755.hg.1 NM_001164665.1 Refseq
Affymetrix Human Clariom D Assay TC0700012755.hg.1 NM_001164665.1 Refseq
Affymetrix Human Clariom D Assay TC0700012755.hg.1 NM_001164665.1 Refseq
Affymetrix Human Clariom D Assay TC0700012755.hg.1 NM_001164665.1 Refseq
Affymetrix Human Clariom D Assay TC0700012755.hg.1 NM_001164665.1 Refseq
Affymetrix Human Clariom D Assay TC0700012755.hg.1 NM_001164665.1 Refseq
Affymetrix Human Clariom D Assay TC0700012755.hg.1 NM_020910.2 Refseq
Affymetrix Human Clariom D Assay TC0700012755.hg.1 NM_020910.2 Refseq
Affymetrix Human Clariom D Assay TC0700012755.hg.1 NM_020910.2 Refseq
Affymetrix Human Clariom D Assay TC0700012755.hg.1 NM_020910.2 Refseq
Affymetrix Human Clariom D Assay TC0700012755.hg.1 NM_020910.2 Refseq
Affymetrix Human Clariom D Assay TC0700012755.hg.1 NM_020910.2 Refseq
Affymetrix Human Clariom D Assay TC0700012755.hg.1 NM_020910.2 Refseq
Affymetrix Human Clariom D Assay TC0700012755.hg.1 NM_020910.2 Refseq
Affymetrix Human Clariom D Assay TC0700012755.hg.1 NM_020910.2 Refseq
Affymetrix Human Clariom D Assay TC0700012755.hg.1 NM_020910.2 Refseq
Affymetrix Human Clariom D Assay TC0700012755.hg.1 NM_020910.2 Refseq
Affymetrix Human Clariom D Assay TC0700012755.hg.1 NM_020910.2 Refseq
Affymetrix Human Clariom D Assay TC0700012755.hg.1 NM_020910.2 Refseq
Affymetrix Human Clariom D Assay TC0700012755.hg.1 NM_020910.2 Refseq
Affymetrix Human Clariom D Assay TC0700012755.hg.1 NM_020910.2 Refseq
Affymetrix Human Clariom D Assay TC0700012755.hg.1 NM_020910.2 Refseq
Affymetrix Human Clariom D Assay TC0700012755.hg.1 NM_020910.2 Refseq
Affymetrix Human Clariom D Assay TC0700012755.hg.1 NM_020910.2 Refseq
Affymetrix Human Clariom D Assay TC0700012755.hg.1 NM_020910.2 Refseq
Affymetrix Human Clariom D Assay TC0700012755.hg.1 NM_020910.2 Refseq
Affymetrix Human Clariom D Assay TC0700012755.hg.1 NM_020910.2 Refseq
Affymetrix Human Clariom D Assay TC0700012755.hg.1 NM_020910.2 Refseq
Affymetrix Human Clariom D Assay TC0700012755.hg.1 NM_020910.2 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-55586bc97b-lttpc:80/100.66.78.80:80.
git-commit: d8c775737291b1e1a3c617895b4ca460ad723db2
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.1-2025.09.37-1.0