This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs05275826_s1
See other EYA3 GE Assays ›
Gene Symbol:
EYA3
Gene Name:
EYA transcriptional coactivator and phosphatase 3
Gene Aliases:
-
Chromosome Location:
Chr.1: 27970344 - 28088671 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs05275826_s1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: EYA3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001282561.1 NP_001269490.1 17 5114 71
NM_001282562.1 NP_001269491.1 17 5172 71
NM_001990.3 NP_001981.2 18 5252 71
XM_006710449.3 XP_006710512.1 19 5288 71
XM_011541004.2 XP_011539306.1 18 5148 71
GenBank mRNA BX648945.1 - 18 5218 71

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 3' Most Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene EYA3

Gene Ontology Categories:

Function(s) Process(es)

double-strand break repair
transcription, DNA-templated
regulation of transcription, DNA-templated
multicellular organism development
visual perception
anatomical structure morphogenesis
response to ionizing radiation
histone dephosphorylation
peptidyl-tyrosine dephosphorylation
positive regulation of DNA repair
protein tyrosine phosphatase activity
protein binding
metal ion binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-kt9qp:80/100.66.76.204:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0