This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs05276106_s1
See other SCP2 GE Assays ›
Gene Symbol:
SCP2
Gene Name:
sterol carrier protein 2
Gene Aliases:
NLTP, NSL-TP, SCP-2, SCP-CHI, SCP-X, SCPX
Chromosome Location:
Chr.1: 52927229 - 53051617 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs05276106_s1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: SCP2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_017002045.1 XP_016857534.1 12 4961 135

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Panther Classification:

Molecular Function -

acyltransferase

Gene Ontology Categories:

Function(s) Process(es)

bile acid biosynthetic process
peroxisome organization
sterol transport
positive regulation of intracellular cholesterol transport
fatty acid beta-oxidation using acyl-CoA oxidase
alpha-linolenic acid metabolic process
lipid hydroperoxide transport
fatty-acyl-CoA binding
receptor binding
protein binding
sterol transporter activity
cholesterol binding
propanoyl-CoA C-acyltransferase activity
long-chain fatty acyl-CoA binding
propionyl-CoA C2-trimethyltridecanoyltransferase activity
oleic acid binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-rpjph:80/100.66.75.107:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0