This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs05278249_g1
See other LPGAT1 GE Assays ›
Gene Symbol:
LPGAT1
Gene Name:
lysophosphatidylglycerol acyltransferase 1
Gene Aliases:
FAM34A, FAM34A1, NET8
Chromosome Location:
Chr.1: 211743457 - 211830772 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs05278249_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LPGAT1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001320808.1 NP_001307737.1 8 6587 109
NM_014873.2 NP_055688.1 8 5998 109
XM_005273362.3 XP_005273419.1 9 6146 109
XM_011510228.2 XP_011508530.1 9 6035 109
XM_011510229.2 XP_011508531.1 8 5918 109
XM_017003004.1 XP_016858493.1 7 7150 109
GenBank mRNA D86960.1 - 8 5999 109

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene LPGAT1

Panther Classification:

Molecular Function -

acyltransferase

Gene Ontology Categories:

Function(s) Process(es)

biological_process
phospholipid biosynthetic process
phosphatidylglycerol acyl-chain remodeling
molecular_function
1-acylglycerol-3-phosphate O-acyltransferase activity
2-acylglycerol-3-phosphate O-acyltransferase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-8cb8z:80/100.66.72.77:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0