This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs05279873_s1
See other CLYBL GE Assays ›
Gene Symbol:
CLYBL
Gene Name:
citrate lyase beta like
Gene Aliases:
CLB
Chromosome Location:
Chr.13: 99606664 - 99909459 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs05279873_s1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer:
435.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: CLYBL
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_017020404.1 XP_016875893.1 1 227 88

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the XM_017020404.1 transcripts.

Set Membership:

Amplicon length between 86 and 100 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Panther Classification:

Molecular Function -

lyase

Gene Ontology Categories:

Process(es)

magnesium ion binding
malate synthase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-49tpj:80/100.66.77.222:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline