This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs05281121_g1
See other VOPP1 GE Assays ›
Gene Symbol:
VOPP1
Gene Name:
vesicular, overexpressed in cancer, prosurvival protein 1
Gene Aliases:
ECOP, GASP
Chromosome Location:
Chr.7: 55434406 - 55572520 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs05281121_g1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: VOPP1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001284282.1 NP_001271211.1 5 2295 65
NM_001284283.1 NP_001271212.1 5 2229 65
NM_001284284.1 NP_001271213.1 5 2390 65
NM_001321242.1 NP_001308171.1 5 2310 65
NM_001321243.1 NP_001308172.1 6 2463 65
NM_001321244.1 NP_001308173.1 6 2496 65
NM_001321246.1 NP_001308175.1 4 2346 65
NM_001321247.1 NP_001308176.1 4 2230 65
NM_001321248.1 NP_001308177.1 4 2173 65
NM_001321249.1 NP_001308178.1 5 2312 65
NM_001321250.1 NP_001308179.1 5 2334 65
NM_001321251.1 NP_001308180.1 5 2320 65
NM_030796.4 NP_110423.3 5 2424 65
GenBank mRNA AB097004.1 - 5 2283 65
AF395824.1 - 4 2189 65
AK000539.1 - 1 584 65
AK075391.1 - 6 2358 65
AK090825.1 - 5 2334 65
AK092992.1 - 5 2368 65
AK126848.1 - 1 2725 65
AL136610.1 - 1 2237 65
BC016650.1 - 1 2316 4342536 65

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene VOPP1

Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription, DNA-templated
signal transduction
signal transducer activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-744bc48644-nrcn4:80/100.66.78.247:80.
git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.48.0-Offline