This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs05296856_s1
See other EXTL2 GE Assays ›
Gene Symbol:
EXTL2
Gene Name:
exostosin like glycosyltransferase 2
Gene Aliases:
EXTR2
Chromosome Location:
Chr.1: 100872372 - 100895411 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs05296856_s1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: EXTL2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001033025.2 NP_001028197.1 1 131 84
NM_001261440.1 NP_001248369.1 1 131 84
NM_001261441.1 NP_001248370.1 1 131 84
NM_001261442.1 NP_001248371.1 1 131 84
NM_001439.3 NP_001430.1 1 131 84
NR_048570.1 1 131 84
XM_005270621.1 XP_005270678.1 1 104 84
XM_011540994.2 XP_011539296.1 1 363 84
XM_011540995.2 XP_011539297.1 1 363 84
GenBank mRNA BC045681.1 - 1 950 5300220 84

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the BC045681.1, NM_001033025.2, NM_001261440.1, NM_001261441.1, NM_001261442.1, NM_001439.3, XM_005270621.1, XM_011540994.2, XM_011540995.2 transcripts.

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene EXTL2

Panther Classification:

Molecular Function -

glycosyltransferase

Gene Ontology Categories:

Function(s) Process(es)

glycosaminoglycan biosynthetic process
N-acetylglucosamine metabolic process
heparan sulfate proteoglycan biosynthetic process
UDP-N-acetylgalactosamine metabolic process
glucuronyl-galactosyl-proteoglycan 4-alpha-N-acetylglucosaminyltransferase activity
alpha-1,4-N-acetylgalactosaminyltransferase activity
metal ion binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-8rxhs:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0