This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs05304798_s1
See other MRPS11 GE Assays ›
Gene Symbol:
MRPS11
Gene Name:
mitochondrial ribosomal protein S11
Gene Aliases:
HCC-2, MRP-S11, S11mt
Chromosome Location:
Chr.15: 88467453 - 88480767 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs05304798_s1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: MRPS11
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001321970.1 NP_001308899.1 6 2225 73
NM_001321972.1 NP_001308901.1 6 3349 73
NM_022839.4 NP_073750.2 6 2228 73
NM_176805.3 NP_789775.1 5 2129 73
NR_135915.1 5 2209 73
NR_135917.1 6 2208 73
NR_135918.1 6 2166 73

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 3' Most Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene MRPS11

Panther Classification:

Molecular Function -

ribosomal protein

Gene Ontology Categories:

Function(s) Process(es)

ribosomal small subunit assembly
maturation of SSU-rRNA from tricistronic rRNA transcript (SSU-rRNA, 5.8S rRNA, LSU-rRNA)
translation
mitochondrial translation
DNA damage response, detection of DNA damage
mitochondrial translational elongation
mitochondrial translational termination
structural constituent of ribosome
poly(A) RNA binding
mRNA 5'-UTR binding
small ribosomal subunit rRNA binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-ng27p:80/100.66.78.145:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline