This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs05312133_s1
See other LMF1 GE Assays ›
Gene Symbol:
LMF1
Gene Name:
lipase maturation factor 1
Gene Aliases:
C16orf26, HMFN1876, JFP11, TMEM112, TMEM112A
Chromosome Location:
Chr.16: 853632 - 981615 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs05312133_s1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LMF1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_017023575.1 XP_016879064.1 11 2259 81

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Gene Ontology Categories:

Function(s)

triglyceride metabolic process
ER to Golgi vesicle-mediated transport
protein secretion
protein glycosylation in Golgi
chylomicron remnant clearance
lipid digestion
positive regulation of lipoprotein lipase activity
protein maturation
regulation of cholesterol metabolic process
regulation of triglyceride metabolic process

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-rr9bx:80/100.66.76.204:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0