This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs05317867_g1
See other PDPR GE Assays ›
Gene Symbol:
PDPR
Gene Name:
pyruvate dehydrogenase phosphatase regulatory subunit
Gene Aliases:
PDP3
Chromosome Location:
Chr.16: 70113626 - 70163630 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs05317867_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PDPR
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_005256016.3 XP_005256073.1 1 346 98

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the XM_005256016.3 transcripts.

Set Membership:

Amplicon length between 86 and 100 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Panther Classification:

Molecular Function -

methyltransferase

Gene Ontology Categories:

Function(s) Process(es)

regulation of acetyl-CoA biosynthetic process from pyruvate
dephosphorylation
oxidation-reduction process
[pyruvate dehydrogenase (lipoamide)] phosphatase activity
oxidoreductase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-9hc65:80/100.66.78.230:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0