This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs05318650_s1
See other WWOX GE Assays ›
Gene Symbol:
WWOX
Gene Name:
WW domain containing oxidoreductase
Gene Aliases:
D16S432E, EIEE28, FOR, FRA16D, HHCMA56, PRO0128, SCAR12, SDR41C1, WOX1
Chromosome Location:
Chr.16: 78099413 - 79212667 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs05318650_s1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: WWOX
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_011523104.2 XP_011521406.1 9 2372 79

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 3' Most Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Panther Classification:

Molecular Function -

transcription cofactor

Gene Ontology Categories:

Function(s) Process(es)

osteoblast differentiation
steroid metabolic process
Wnt signaling pathway
negative regulation of Wnt signaling pathway
positive regulation of transcription from RNA polymerase II promoter
skeletal system morphogenesis
oxidation-reduction process
cellular response to transforming growth factor beta stimulus
intrinsic apoptotic signaling pathway by p53 class mediator
extrinsic apoptotic signaling pathway
positive regulation of extrinsic apoptotic signaling pathway
positive regulation of extrinsic apoptotic signaling pathway in absence of ligand
RNA polymerase II transcription coactivator activity
protein binding
oxidoreductase activity
enzyme binding
protein dimerization activity
cofactor binding
coenzyme binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-fcf5f9d6d-9k2pj:80/100.66.79.197:80.
git-commit: 856de8fa7eff2d55603d92afdf035972e45c3e8a
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.51.0-2026.05.21-1.0