This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs05331285_gH
See other RPL13 GE Assays ›
Gene Symbol:
RPL13
Gene Name:
ribosomal protein L13
Gene Aliases:
BBC1, D16S444E, D16S44E, L13
Chromosome Location:
Chr.16: 89560657 - 89566829 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs05331285_gH
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: RPL13
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_000977.3 NP_000968.2 4 398 80
NM_001243130.1 NP_001230059.1 4 341 80
NM_033251.2 NP_150254.1 3 571 80
GenBank mRNA AB062392.1 - 3 347 80
AK025405.1 - 1 392 80
AK026501.1 - 1 992 80
AK127579.1 - 4 375 80
AK130963.1 - 4 294 80
AK130964.1 - 4 243 80
AK290893.1 - 4 373 80
AK294655.1 - 4 80
AK308545.1 - 3 213 80
BC000851.1 - 1 351 80
BC004954.1 - 1 361 3623320 80
BC006179.1 - 4 80
BC007345.2 - 4 363 3688320 80
BC007563.1 - 3 559 2989021 80
BC007805.2 - 4 359 4298327 80
BC010994.2 - 3 354 3842093 80
BC013078.1 - 4 359 3459415 80
BC014167.2 - 1 359 4550361 80
BC020804.1 - 3 571 4276103 80
BC027463.1 - 4 359 5111302 80
BC063378.1 - 1 362 3542514 80
BC066320.1 - 4 373 5300601 80
BC093063.1 - 1 30401947 80
BC106058.1 - 4 398 4992467 80
BG706207.1 - 4 320 80
X64707.1 - 4 80

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene RPL13

Panther Classification:

Molecular Function -

ribosomal protein

Gene Ontology Categories:

Function(s) Process(es)

nuclear-transcribed mRNA catabolic process, nonsense-mediated decay
rRNA processing
translation
translational initiation
SRP-dependent cotranslational protein targeting to membrane
viral transcription
RNA binding
structural constituent of ribosome
protein binding
poly(A) RNA binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-zvjvc:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline