This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs05337056_s1
See other NPC1 GE Assays ›
Gene Symbol:
NPC1
Gene Name:
NPC intracellular cholesterol transporter 1
Gene Aliases:
NPC
Chromosome Location:
Chr.18: 23506184 - 23586617 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs05337056_s1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: NPC1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_006722479.3 XP_006722542.1 25 4570 108
XM_017025787.1 XP_016881276.1 25 4519 108

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Gene Ontology Categories:

Function(s) Process(es)

protein glycosylation
endocytosis
autophagy
lysosomal transport
signal transduction
adult walking behavior
cholesterol metabolic process
bile acid metabolic process
negative regulation of macroautophagy
cholesterol transport
membrane raft organization
cholesterol efflux
response to drug
cholesterol homeostasis
response to cadmium ion
viral entry into host cell
negative regulation of cell death
cellular response to steroid hormone stimulus
cellular response to low-density lipoprotein particle stimulus
establishment of protein localization to membrane
positive regulation of cholesterol homeostasis
virus receptor activity
receptor activity
transmembrane signaling receptor activity
protein binding
sterol transporter activity
cholesterol binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-49tpj:80/100.66.77.222:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline