This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs05342054_s1
See other LIPG GE Assays ›
Gene Symbol:
LIPG
Gene Name:
lipase G, endothelial type
Gene Aliases:
EDL, EL, PRO719
Chromosome Location:
Chr.18: 49560699 - 49592908 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs05342054_s1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LIPG
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_005258390.1 XP_005258447.1 1 853 70
XM_011526265.2 XP_011524567.1 1 66 70
GenBank mRNA AK125344.1 - 1 853 70

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the XM_005258390.1, XM_011526265.2 transcripts.

Set Membership:

Amplicon length less than or equal to 70 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Panther Classification:

Molecular Function -

lipase

Gene Ontology Categories:

Function(s) Process(es)

lipid metabolic process
response to nutrient
cell proliferation
phospholipid catabolic process
positive regulation of high-density lipoprotein particle clearance
positive regulation of cholesterol transport
high-density lipoprotein particle remodeling
cholesterol homeostasis
reverse cholesterol transport
lipid digestion
regulation of lipoprotein metabolic process
phospholipid homeostasis
lipoprotein lipase activity
phospholipase activity
heparin binding
phosphatidylcholine 1-acylhydrolase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0