This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs05354320_s1
See other CIC GE Assays ›
Gene Symbol:
CIC
Gene Name:
capicua transcriptional repressor
Gene Aliases:
-
Chromosome Location:
Chr.19: 42268537 - 42295796 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs05354320_s1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: CIC
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001304815.1 NP_001291744.1 2 2032 94
XM_011526660.2 XP_011524962.1 2 2074 94
XM_011526661.2 XP_011524963.1 2 2074 94
XM_011526662.2 XP_011524964.1 2 2074 94
XM_011526663.2 XP_011524965.1 2 2074 94
XM_011526664.2 XP_011524966.1 2 2074 94
XM_011526665.2 XP_011524967.1 2 2074 94
XM_011526666.2 XP_011524968.1 2 2074 94
XM_011526667.2 XP_011524969.1 2 2074 94

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Panther Classification:

Molecular Function -

DNA-binding transcription factor

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of transcription from RNA polymerase II promoter
transcription, DNA-templated
lung alveolus development
DNA binding
chromatin binding
protein binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mt7zw:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0