This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs05354592_m1
See other KPTN GE Assays ›
Gene Symbol:
KPTN
Gene Name:
kaptin, actin binding protein
Gene Aliases:
2E4, MRT41
Chromosome Location:
Chr.19: 47475141 - 47486792 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs05354592_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: KPTN
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_017026226.1 XP_016881715.1 2 - 3 356 110
XM_017026227.1 XP_016881716.1 1 - 2 432 110
XM_017026228.1 XP_016881717.1 2 - 3 356 110
XM_017026229.1 XP_016881718.1 2 - 3 356 110

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Homo sapiens Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

movement of cell or subcellular component
actin filament organization
sensory perception of sound
actin binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-hhcgv:80/100.66.79.225:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0