This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs05355993_s1
See other MBOAT2 GE Assays ›
Gene Symbol:
MBOAT2
Gene Name:
membrane bound O-acyltransferase domain containing 2
Gene Aliases:
LPAAT, LPCAT4, LPEAT, LPLAT 2, OACT2
Chromosome Location:
Chr.2: 8852690 - 9003750 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs05355993_s1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: MBOAT2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001321265.1 NP_001308194.1 12 4849 77
NM_001321266.1 NP_001308195.1 11 4771 77
NM_001321267.1 NP_001308196.1 14 5072 77
NM_138799.3 NP_620154.2 13 4995 77
NR_135598.1 14 5047 77
NR_135599.1 13 5314 77
NR_135600.1 13 4901 77
NR_135601.1 11 4690 77
NR_135602.1 12 4862 77
NR_135603.1 14 5088 77
NR_135604.1 13 5011 77
NR_135605.1 12 4823 77
NR_135606.1 13 4914 77

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene MBOAT2

Panther Classification:

Molecular Function -

acetyltransferase

Gene Ontology Categories:

Function(s) Process(es)

phospholipid biosynthetic process
phosphatidylcholine acyl-chain remodeling
phosphatidylethanolamine acyl-chain remodeling
1-acylglycerol-3-phosphate O-acyltransferase activity
2-acylglycerol-3-phosphate O-acyltransferase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-hk65g:80/100.66.77.69:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0