This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs05357875_s1
See other CDKL4 GE Assays ›
Gene Symbol:
CDKL4
Gene Name:
cyclin dependent kinase like 4
Gene Aliases:
-
Chromosome Location:
Chr.2: 39174421 - 39246815 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs05357875_s1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: CDKL4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_011532815.2 XP_011531117.1 1 685 66
XM_011532816.2 XP_011531118.1 1 685 66
XM_011532817.2 XP_011531119.1 1 685 66
XM_011532819.2 XP_011531121.1 1 685 66
XM_017003983.1 XP_016859472.1 1 685 66
XM_017003984.1 XP_016859473.1 1 685 66
XM_017003985.1 XP_016859474.1 1 685 66

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the XM_011532815.2, XM_011532816.2, XM_011532817.2, XM_011532819.2, XM_017003983.1, XM_017003984.1, XM_017003985.1 transcripts.

Set Membership:

Amplicon length less than or equal to 70 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Panther Classification:

Molecular Function -

non-receptor serine/threonine protein kinase

Gene Ontology Categories:

Function(s) Process(es)

protein phosphorylation
regulation of cell cycle
cyclin-dependent protein serine/threonine kinase activity
ATP binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-zvjvc:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline