This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs05375712_g1
See other EIF3A GE Assays ›
Gene Symbol:
EIF3A
Gene Name:
eukaryotic translation initiation factor 3 subunit A
Gene Aliases:
EIF3, EIF3S10, P167, TIF32, eIF3-p170, eIF3-theta, p180, p185
Chromosome Location:
Chr.10: 119035029 - 119080822 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs05375712_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: EIF3A
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_003750.2 NP_003741.1 22 4971 92
GenBank mRNA AJ227915.1 - 1 109 92
BC020285.1 - 4 1390 92
D50929.1 - 22 4954 92
U58046.1 - 22 4922 92
U78311.1 - 22 4937 92

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene EIF3A

Panther Classification:

Molecular Function -

translation factor translation initiation factor translational protein

Gene Ontology Categories:

Function(s) Process(es)

formation of translation preinitiation complex
formation of cytoplasmic translation initiation complex
translational initiation
regulation of translational initiation
IRES-dependent viral translational initiation
viral translational termination-reinitiation
translation initiation factor activity
structural molecule activity
protein binding
poly(A) RNA binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-744bc48644-5wn4c:80/100.66.76.150:80.
git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.48.0-Offline