This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs05394251_s1
See other ERG GE Assays ›
Gene Symbol:
ERG
Gene Name:
ERG, ETS transcription factor
Gene Aliases:
erg-3, p55
Chromosome Location:
Chr.21: 38367261 - 38662045 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs05394251_s1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ERG
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001136154.1 NP_001129626.1 12 2334 93
NM_001136155.1 NP_001129627.1 9 1947 93
NM_001243428.1 NP_001230357.1 12 2357 93
NM_001243429.1 NP_001230358.1 9 1921 93
NM_004449.4 NP_004440.1 11 2262 93
NM_182918.3 NP_891548.1 10 2165 93
XM_011529486.2 XP_011527788.1 10 2444 93
XM_017028287.1 XP_016883776.1 11 2518 93
XM_017028288.1 XP_016883777.1 9 2047 93
XM_017028289.1 XP_016883778.1 8 1966 93

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 3' Most Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene ERG

Panther Classification:

Molecular Function -

winged helix/forkhead transcription factor

Gene Ontology Categories:

Function(s) Process(es)

endocardial cushion development
endocardial cushion to mesenchymal transition involved in heart valve formation
transcription, DNA-templated
regulation of transcription from RNA polymerase II promoter
protein phosphorylation
signal transduction
multicellular organism development
cell proliferation
cell migration
cell differentiation
positive regulation of blood vessel remodeling
RNA polymerase II transcription factor activity, sequence-specific DNA binding
DNA binding
chromatin binding
signal transducer activity
protein binding
sequence-specific DNA binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0