This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs05397430_s1
See other PRMT2 GE Assays ›
Gene Symbol:
PRMT2
Gene Name:
protein arginine methyltransferase 2
Gene Aliases:
HRMT1L1
Chromosome Location:
Chr.21: 46635153 - 46665685 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs05397430_s1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PRMT2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001242866.2 NP_001229795.1 7 2443 78
GenBank mRNA BY799824.2 - 1 234 78

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 3' Most Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Panther Classification:

Molecular Function -

protein modifying enzyme

Gene Ontology Categories:

Function(s) Process(es)

protein methylation
signal transduction
histone methylation
peptidyl-arginine methylation, to asymmetrical-dimethyl arginine
negative regulation of NF-kappaB transcription factor activity
histone arginine methylation
positive regulation of apoptotic process
negative regulation of transcription, DNA-templated
positive regulation of transcription, DNA-templated
developmental cell growth
regulation of androgen receptor signaling pathway
negative regulation of G1/S transition of mitotic cell cycle
transcription coactivator activity
signal transducer activity
protein binding
histone-arginine N-methyltransferase activity
protein-arginine N-methyltransferase activity
estrogen receptor binding
progesterone receptor binding
protein-arginine omega-N asymmetric methyltransferase activity
histone methyltransferase activity
protein homodimerization activity
retinoic acid receptor binding
peroxisome proliferator activated receptor binding
thyroid hormone receptor binding
androgen receptor binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mvcqg:80/100.66.75.199:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0