This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs05407983_s1
See other SCAP GE Assays ›
Gene Symbol:
SCAP
Gene Name:
SREBF chaperone
Gene Aliases:
-
Chromosome Location:
Chr.3: 47413685 - 47477126 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs05407983_s1
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: SCAP
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001320044.1 NP_001306973.1 9 1278 55
NM_012235.3 NP_036367.2 12 1763 55
XM_005264967.1 XP_005265024.1 12 1690 55
XM_005264968.1 XP_005265025.1 9 1432 55
XM_005264971.1 XP_005265028.1 6 843 55
XM_011533501.1 XP_011531803.1 12 1790 55
XM_011533502.2 XP_011531804.1 4 439 55
XM_017005918.1 XP_016861407.1 12 1777 55
XM_017005919.1 XP_016861408.1 12 1710 55
XM_017005920.1 XP_016861409.1 12 1710 55
XM_017005921.1 XP_016861410.1 6 843 55
GenBank mRNA AK027402.1 - 6 756 55
AK291484.1 - 7 956 55
AK300311.1 - 9 1168 55
BC020987.1 - 4 484 3852258 55
BC027207.2 - 8 962 55
D83782.2 - 12 1749 55

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene SCAP

Gene Ontology Categories:

Function(s) Process(es)

response to hypoxia
aging
cholesterol metabolic process
response to insulin
SREBP signaling pathway
regulation of fatty acid biosynthetic process
negative regulation of cholesterol biosynthetic process
positive regulation of low-density lipoprotein particle receptor biosynthetic process
protein binding
cholesterol binding
protein complex binding
unfolded protein binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-zpsr4:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0