This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs05412152_g1
See other CCT8 GE Assays ›
Gene Symbol:
CCT8 FMO5
Gene Name:
chaperonin containing TCP1 subunit 8
flavin containing monooxygenase 5
Gene Aliases:
C21orf112, Cctq, D21S246, PRED71
-
Chromosome Location:
Chr.21: 29056322 - 29073797 on Build GRCh38
Chromosome Location:
Chr.1: 147183963 - 147225798 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
-
Assay ID Hs05412152_g1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: CCT8
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001282907.1 NP_001269836.1 8 1236 106
NM_001282908.1 NP_001269837.1 7 953 106
NM_001282909.1 NP_001269838.1 6 848 106
NM_006585.3 NP_006576.2 7 928 106
GenBank mRNA AK222490.1 - 7 746 106
AK293705.1 - 8 790 106
AK298801.1 - 6 702 106
AK308063.1 - 7 780 106
BC012584.1 - 7 795 4133182 106
BC035472.1 - 7 744 106
BC072001.1 - 7 786 6298609 106
BC095470.1 - 7 768 30523886 106
D13627.1 - 7 749 106
Gene Symbol: FMO5
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_006711244.3 XP_006711307.1 7 2006 105

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene CCT8
Click here to find siRNAs for Gene FMO5

Panther Classification:

Molecular Function -

chaperonin oxidoreductase oxygenase metabolite interconversion enzyme

Gene Ontology Categories:

Function(s) Process(es)

protein folding
binding of sperm to zona pellucida
positive regulation of telomere maintenance via telomerase
pore complex assembly
protein stabilization
cell-cell adhesion
toxin transport
positive regulation of establishment of protein localization to telomere
positive regulation of protein localization to Cajal body
positive regulation of telomerase RNA localization to Cajal body
biological_process
drug metabolic process
oxidation-reduction process
protein binding
ATP binding
ATPase activity, coupled
unfolded protein binding
cadherin binding involved in cell-cell adhesion
N,N-dimethylaniline monooxygenase activity
flavin adenine dinucleotide binding
NADP binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-744bc48644-fvx7n:80/100.66.78.247:80.
git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.48.0-Offline