Assay Details
| Gene Symbol: LOC653513 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017003021.1 | XP_016858510.1 | 8 | 2633 | 152 | |
| XM_017003023.1 | XP_016858512.1 | 7 | 2522 | 152 | ||
| XM_017003024.1 | XP_016858513.1 | 8 | 4635 | 152 | ||
| XM_017003025.1 | XP_016858514.1 | 8 | 4375 | 152 | ||
| XM_017003027.1 | XP_016858516.1 | 8 | 2468 | 152 | ||
| XM_017003028.1 | XP_016858517.1 | 7 | 4524 | 152 | ||
| XM_017003029.1 | XP_016858518.1 | 7 | 4264 | 152 | ||
| XM_017003030.1 | XP_016858519.1 | 7 | 2364 | 152 | ||
| XM_017003031.1 | XP_016858520.1 | 8 | 4470 | 152 | ||
| XM_017003032.1 | XP_016858521.1 | 8 | 4210 | 152 | ||
| XM_017003036.1 | XP_016858525.1 | 7 | 4366 | 152 | ||
| XM_017003037.1 | XP_016858526.1 | 7 | 4106 | 152 | ||
| XM_017003038.1 | XP_016858527.1 | 5 | 3243 | 152 | ||
More Information
Set Membership: |
|

LOC653513
Genomic Map