This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs05424251_s1
See other NME6 GE Assays ›
Gene Symbol:
NME6 FCF1P2
Gene Name:
NME/NM23 nucleoside diphosphate kinase 6
FCF1 pseudogene 2
Gene Aliases:
IPIA-ALPHA, NDK 6, NM23-H6
-
Chromosome Location:
Chr.3: 48289369 - 48302259 on Build GRCh38
Chromosome Location:
Chr.3: 48290279 - 48291678 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
-
Assay ID Hs05424251_s1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: NME6
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_017005512.1 XP_016861001.1 3 4915 80
XM_017005513.1 XP_016861002.1 6 4306 80
XM_017005514.1 XP_016861003.1 7 4563 80
XM_017005515.1 XP_016861004.1 5 4319 80
XM_017005517.1 XP_016861006.1 3 4081 80
XM_017005518.1 XP_016861007.1 4 4210 80
Gene Symbol: FCF1P2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_134638.1 1 722 80
GenBank mRNA CU451957.1 - 1 215 80

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Panther Classification:

Molecular Function -

kinase transferase metabolite interconversion enzyme

Gene Ontology Categories:

Function(s) Process(es)

nucleoside diphosphate phosphorylation
GTP biosynthetic process
UTP biosynthetic process
CTP biosynthetic process
apoptotic process
negative regulation of cell growth
negative regulation of mitotic nuclear division
nucleoside diphosphate kinase activity
ATP binding
metal ion binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-744bc48644-454p7:80/100.66.72.101:80.
git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.48.0-Offline