This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs05429431_gH
See other METTL15 GE Assays ›
Gene Symbol:
METTL15
Gene Name:
methyltransferase like 15
Gene Aliases:
METT5D1
Chromosome Location:
Chr.11: 28108251 - 28527041 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs05429431_gH
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: METTL15
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001113528.1 NP_001107000.1 7 1453 105
NM_001297775.1 NP_001284704.1 7 1409 105
NM_152636.2 NP_689849.2 8 1524 105
XM_011519935.2 XP_011518237.1 9 1674 105
XM_011519937.2 XP_011518239.1 6 915 105
XM_011519940.2 XP_011518242.1 6 1274 105
XM_011519941.2 XP_011518243.1 6 1217 105
XM_017017297.1 XP_016872786.1 7 1357 105
XM_017017298.1 XP_016872787.1 10 1713 105
XM_017017299.1 XP_016872788.1 6 2202 105
XM_017017300.1 XP_016872789.1 8 1410 105
XM_017017302.1 XP_016872791.1 5 812 105
XM_017017303.1 XP_016872792.1 9 1612 105
GenBank mRNA AK091298.1 - 7 1176 105
AL832668.1 - 6 1106 105
BC030997.1 - 6 1082 4657323 105
BC066900.1 - 7 1291 105

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene METTL15

Panther Classification:

Molecular Function -

methyltransferase

Gene Ontology Categories:

Function(s) Process(es)

rRNA base methylation
rRNA (cytosine-N4-)-methyltransferase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-wt24k:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline