This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs05454370_g1
See other CDH12 GE Assays ›
Gene Symbol:
CDH12
Gene Name:
cadherin 12
Gene Aliases:
CDHB
Chromosome Location:
Chr.5: 21750191 - 22853622 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs05454370_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: CDH12
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001317227.1 NP_001304156.1 3 804 111
NM_001317228.1 NP_001304157.1 4 919 111
NM_004061.4 NP_004052.2 6 1433 111
XM_011513927.2 XP_011512229.1 4 710 111
XM_017008920.1 XP_016864409.1 5 1060 111
XM_017008921.1 XP_016864410.1 6 1153 111
GenBank mRNA AK295123.1 - 4 919 111
AK314800.1 - 3 804 111
BC047608.1 - 6 1138 5751948 111
L33477.1 - 6 1434 111
L34057.1 - 2 439 111

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene CDH12

Panther Classification:

Molecular Function -

cadherin

Gene Ontology Categories:

Function(s) Process(es)

homophilic cell adhesion via plasma membrane adhesion molecules
adherens junction organization
calcium ion binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-55658555d6-9rjdb:80/100.66.79.224:80.
git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.41.0-Offline