This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs05469653_s1
See other GABRA1 GE Assays ›
Gene Symbol:
GABRA1
Gene Name:
gamma-aminobutyric acid type A receptor alpha1 subunit
Gene Aliases:
ECA4, EIEE19, EJM, EJM5
Chromosome Location:
Chr.5: 161847191 - 161899959 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs05469653_s1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer:
435.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: GABRA1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_000806.5 NP_000797.2 11 4266 78
NM_001127643.1 NP_001121115.1 11 4153 78
NM_001127644.1 NP_001121116.1 10 4302 78
NM_001127645.1 NP_001121117.1 10 3890 78
NM_001127648.1 NP_001121120.1 9 3912 78

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 3' Most Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene GABRA1

Panther Classification:

Molecular Function -

ligand-gated ion channel

Gene Ontology Categories:

Function(s) Process(es)

transport
gamma-aminobutyric acid signaling pathway
ion transmembrane transport
synaptic transmission, GABAergic
cellular response to histamine
chloride transmembrane transport
GABA-A receptor activity
drug binding
GABA receptor activity
GABA-gated chloride ion channel activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-49tpj:80/100.66.77.222:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline