This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs05482521_s1
See other PHACTR1 GE Assays ›
Gene Symbol:
PHACTR1
Gene Name:
phosphatase and actin regulator 1
Gene Aliases:
RPEL, RPEL1, dJ257A7.2
Chromosome Location:
Chr.6: 12716635 - 13290484 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs05482521_s1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer:
435.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PHACTR1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_005248934.2 XP_005248991.1 16 5305 97
XM_017010452.1 XP_016865941.1 12 5809 97
XM_017010454.1 XP_016865943.1 15 6145 97
XM_017010455.1 XP_016865944.1 15 6113 97
XM_017010456.1 XP_016865945.1 14 6110 97
XM_017010457.1 XP_016865946.1 14 6089 97
XM_017010458.1 XP_016865947.1 11 5602 97
XM_017010459.1 XP_016865948.1 14 5938 97
XM_017010460.1 XP_016865949.1 14 5906 97
XM_017010461.1 XP_016865950.1 13 5877 97
XM_017010462.1 XP_016865951.1 13 5903 97
XM_017010463.1 XP_016865952.1 12 6003 97
XM_017010464.1 XP_016865953.1 12 4762 97
XM_017010465.1 XP_016865954.1 12 5783 97
XM_017010466.1 XP_016865955.1 12 5796 97
XM_017010467.1 XP_016865956.1 11 5783 97
XM_017010469.1 XP_016865958.1 11 5585 97
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA AK024681.1 - 1 Chr. 6 - 13288131 - 13290482 [-] on Build GRCh38 97

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 3' Most Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Panther Classification:

Molecular Function -

phosphatase modulator

Gene Ontology Categories:

Function(s) Process(es)

actomyosin structure organization
actin cytoskeleton reorganization
negative regulation of catalytic activity
stress fiber assembly
cell motility
actin binding
protein phosphatase inhibitor activity
protein phosphatase 1 binding
protein phosphatase type 1 regulator activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-49tpj:80/100.66.77.222:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline