This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs05490039_s1
See other SHPRH GE Assays ›
Gene Symbol:
SHPRH
Gene Name:
SNF2 histone linker PHD RING helicase
Gene Aliases:
bA545I5.2
Chromosome Location:
Chr.6: 145857838 - 145964436 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs05490039_s1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: SHPRH
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_006715439.3 XP_006715502.1 31 11498 100

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 3' Most Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Panther Classification:

Molecular Function -

ubiquitin-protein ligase

Gene Ontology Categories:

Function(s) Process(es)

protein polyubiquitination
DNA repair
nucleosome assembly
cellular response to DNA damage stimulus
DNA binding
helicase activity
ubiquitin-protein transferase activity
protein binding
ATP binding
zinc ion binding
ligase activity
ubiquitin protein ligase binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-z4mpm:80/100.66.76.55:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0