This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs05495069_s1
See other TMED4 GE Assays ›
Gene Symbol:
TMED4
Gene Name:
transmembrane p24 trafficking protein 4
Gene Aliases:
ERS25, GMP25iso, HNLF, p24a3, p24alpha3
Chromosome Location:
Chr.7: 44577894 - 44582295 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs05495069_s1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: TMED4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001303059.1 NP_001289988.1 4 1815 128
NM_001303061.1 NP_001289990.1 5 1955 128
NM_001303062.1 NP_001289991.1 4 2185 128
NM_182547.3 NP_872353.2 5 1962 128
GenBank mRNA BC035467.1 - 5 1886 128

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene TMED4

Panther Classification:

Molecular Function -

vesicle coat protein

Gene Ontology Categories:

Function(s) Process(es)

signal transduction
protein transport
positive regulation of I-kappaB kinase/NF-kappaB signaling
signal transducer activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-l4hk6:80/100.66.76.55:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0