This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs05504262_s1
See other HECW1 GE Assays ›
Gene Symbol:
HECW1
Gene Name:
HECT, C2 and WW domain containing E3 ubiquitin protein ligase 1
Gene Aliases:
NEDL1
Chromosome Location:
Chr.7: 43112580 - 43566001 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs05504262_s1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: HECW1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_005249665.3 XP_005249722.1 29 8472 90
XM_006715670.3 XP_006715733.1 28 8370 90
XM_006715671.3 XP_006715734.1 28 8367 90
XM_006715673.3 XP_006715736.1 27 8265 90
XM_011515220.2 XP_011513522.1 29 8263 90
XM_011515222.2 XP_011513524.1 28 8404 90
XM_011515223.2 XP_011513525.1 28 8205 90
XM_011515224.2 XP_011513526.1 28 8685 90
XM_011515225.2 XP_011513527.2 26 7722 90
XM_011515226.2 XP_011513528.1 25 7946 90
XM_017011882.1 XP_016867371.1 28 8712 90
XM_017011883.1 XP_016867372.1 29 8649 90
XM_017011884.1 XP_016867373.1 28 8709 90
XM_017011885.1 XP_016867374.1 30 8645 90
XM_017011886.1 XP_016867375.1 29 8474 90
XM_017011887.1 XP_016867376.1 27 8610 90
XM_017011888.1 XP_016867377.1 27 8607 90
XM_017011889.1 XP_016867378.1 28 8664 90

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 3' Most Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Panther Classification:

Molecular Function -

ubiquitin-protein ligase

Gene Ontology Categories:

Function(s) Process(es)

protein ubiquitination involved in ubiquitin-dependent protein catabolic process
negative regulation of canonical Wnt signaling pathway
ubiquitin-protein transferase activity
ligase activity
ubiquitin protein ligase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-rr9bx:80/100.66.76.204:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0