This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs05513682_s1
See other TTPA GE Assays ›
Gene Symbol:
TTPA
Gene Name:
alpha tocopherol transfer protein
Gene Aliases:
ATTP, AVED, TTP1, alphaTTP
Chromosome Location:
Chr.8: 63058409 - 63086523 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs05513682_s1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer:
435.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: TTPA
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_006716468.3 XP_006716531.1 3 3414 104

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Panther Classification:

Molecular Function -

transfer/carrier protein

Gene Ontology Categories:

Function(s) Process(es)

embryonic placenta development
lipid metabolic process
transport
response to nutrient
response to pH
response to toxic substance
vitamin E metabolic process
intermembrane transport
vitamin transport
intracellular pH reduction
negative regulation of cell death
negative regulation of establishment of blood-brain barrier
transporter activity
protein binding
phosphatidylinositol-4,5-bisphosphate binding
vitamin E binding
phosphatidylinositol-3,4-bisphosphate binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-zvjvc:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline