This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs05514921_g1
See other CP GE Assays ›
Gene Symbol:
CP HPS3
Gene Name:
ceruloplasmin (ferroxidase)
HPS3, biogenesis of lysosomal organelles complex 2 subunit 1
Gene Aliases:
CP-2
BLOC2S1, SUTAL
Chromosome Location:
Chr.3: 149162410 - 149222050 on Build GRCh38
Chromosome Location:
Chr.3: 149129584 - 149173196 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
-
Assay ID Hs05514921_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: CP
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_000096.3 NP_000087.1 19 3860 86
XM_006713500.3 XP_006713563.1 19 3877 86
XM_017005735.1 XP_016861224.1 18 3702 86
GenBank mRNA BC039239.1 - 8 1504 86
BC146801.1 - 19 3640 86
BX647327.1 - 5 1463 86
Gene Symbol: HPS3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA BG618578.1 - 1 214 86

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene CP
Click here to find siRNAs for Gene HPS3

Panther Classification:

Molecular Function -

oxidase

Gene Ontology Categories:

Function(s) Process(es)

liver development
copper ion transport
cellular iron ion homeostasis
aging
response to nutrient
lung development
response to copper ion
oxidation-reduction process
ferroxidase activity
copper ion binding
chaperone binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-dd86746ff-77ss8:80/100.66.75.30:80.
git-commit: 04e44c58f20fafe66519586ffe6db5d8a90e52ce
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.53.1-Offline