This product has been added to your favorites list. Go to
My Favorites
Please enter the information below and press OK to send your cart to Core Services for purchase.
Name
Phone number
Email
Budget number
PI Name
PI Email
Building and Room number
Pickup
Delivery
System Message
OK
Cancel
LOADING
Home
›
Search Tool
›
Search Results
› Hs05518517_s1
See other DMTN GE Assays ›
Gene Symbol:
DMTN
Gene Name:
dematin actin binding protein
Gene Aliases:
DMT, EPB49
Chromosome Location:
Chr.8: 22048955 - 22082527 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID
Hs05518517_s1
Size
FAM-MGB | XS: 75 rxns
FAM-MGB | S: 360 rxns
FAM-MGB | M: 750 rxns
FAM-MGB | L: 2900 rxns
VIC-MGB_PL | S: 360 rxns
VIC-MGB_PL | M: 750 rxns
VIC-MGB_PL | L: 2900 rxns
VIC-MGB | S: 360 rxns
VIC-MGB | M: 750 rxns
VIC-MGB | L: 2900 rxns
Availability
Made To Order
Catalog #
4351372
Price
435.00
Your Price
Online offer:
408.65
408.65
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Gene Symbol: DMTN
Interrogated Sequence
Translated Protein
Exon Boundary
Assay Location
IMAGE Clone ID
Amplicon Length
RefSeq
NM_001114135.4
NP_001107607.1
16
1826
85
NM_001114136.2
NP_001107608.1
16
1782
85
NM_001114137.3
NP_001107609.1
15
1890
85
NM_001114138.2
NP_001107610.1
15
1877
85
NM_001114139.3
NP_001107611.1
14
1774
85
NM_001302816.2
NP_001289745.1
15
2010
85
NM_001302817.2
NP_001289746.1
14
1795
85
NM_001323378.1
NP_001310307.1
16
1915
85
NM_001323380.1
NP_001310309.1
16
1822
85
NM_001323381.1
NP_001310310.1
15
1931
85
NM_001323382.1
NP_001310311.1
16
1861
85
NM_001323383.1
NP_001310312.1
15
1716
85
NM_001323384.1
NP_001310313.1
15
1849
85
NM_001323385.1
NP_001310314.1
14
1865
85
NM_001323387.1
NP_001310316.1
14
1814
85
NM_001323388.1
NP_001310317.1
14
1662
85
NM_001323389.1
NP_001310318.1
15
1870
85
NM_001323390.1
NP_001310319.1
15
1737
85
NM_001323391.1
NP_001310320.1
15
1840
85
NM_001323392.1
NP_001310321.1
15
2001
85
NM_001323393.1
NP_001310322.1
15
1707
85
NM_001323394.1
NP_001310323.1
15
1868
85
NM_001323395.1
NP_001310324.1
15
1747
85
NM_001323396.1
NP_001310325.1
15
1751
85
NM_001323397.1
NP_001310326.1
15
1919
85
NM_001323398.1
NP_001310327.1
13
1729
85
NM_001323399.1
NP_001310328.1
13
1596
85
NM_001323400.1
NP_001310329.1
12
1745
85
NM_001323401.1
NP_001310330.1
13
1769
85
NM_001978.4
NP_001969.2
16
2076
85
XM_005273432.1
XP_005273489.1
16
1808
85
XM_005273433.1
XP_005273490.1
16
1740
85
XM_005273442.1
XP_005273499.1
15
1592
85
XM_011544435.1
XP_011542737.1
15
2001
85
XM_017013189.1
XP_016868678.1
15
1683
85
XM_017013190.1
XP_016868679.1
14
1935
85
XM_017013191.1
XP_016868680.1
14
1689
85
XM_017013192.1
XP_016868681.1
14
1570
85
XM_017013193.1
XP_016868682.1
14
1768
85
XM_017013194.1
XP_016868683.1
14
1635
85
XM_017013195.1
XP_016868684.1
14
1935
85
XM_017013196.1
XP_016868685.1
14
1538
85
XM_017013197.1
XP_016868686.1
14
1769
85
XM_017013198.1
XP_016868687.1
14
1611
85
GenBank mRNA
AB209718.1
-
12
1302
85
AK055842.1
-
15
1674
85
AK091581.1
-
14
1689
85
BC006318.2
-
16
1825
4125804
85
BC017445.1
-
16
1825
3162732
85
BC052805.1
-
15
1800
6269658
85
BI222385.1
-
1
107
85
L19713.1
-
15
1936
83
U28389.1
-
16
2002
83
Back To Top
More Information
Set Membership:
Ambion siRNA available
Amplicon length between 71 and 85
3' Most
Mammalian Gene Collection
Detect Genomic DNA
Homo sapiens
Both primers and probe map within a single exon
Matching Ambion siRNAs:
Click here to find siRNAs for Gene DMTN
Panther Classification:
Molecular Function -
actin or actin-binding cytoskeletal protein
Gene Ontology Categories:
Function(s)
Process(es)
protein complex assembly
cytoskeleton organization
regulation of cell shape
regulation of lamellipodium assembly
positive regulation of fibroblast migration
negative regulation of peptidyl-threonine phosphorylation
negative regulation of cell-substrate adhesion
actin cytoskeleton organization
positive regulation of blood coagulation
regulation of actin cytoskeleton organization
negative regulation of peptidyl-serine phosphorylation
calcium-mediated signaling using intracellular calcium source
calcium-mediated signaling using extracellular calcium source
erythrocyte development
negative regulation of peptidyl-tyrosine phosphorylation
actin filament bundle assembly
regulation of filopodium assembly
actin filament capping
negative regulation of focal adhesion assembly
transmembrane transport
protein secretion by platelet
cellular response to calcium ion
cellular response to cAMP
positive regulation of wound healing
negative regulation of protein targeting to membrane
actin filament reorganization
negative regulation of substrate adhesion-dependent cell spreading
positive regulation of substrate adhesion-dependent cell spreading
positive regulation of platelet aggregation
positive regulation of integrin-mediated signaling pathway
actin binding
receptor binding
protein binding
spectrin binding
Back To Top
Related Products
TaqMan
®
Fast Advanced Master Mix
SuperScript
®
VILO
™
cDNA Synthesis Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-mt7zw:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0