This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs05523631_s1
See other TRPS1 GE Assays ›
Gene Symbol:
TRPS1
Gene Name:
transcriptional repressor GATA binding 1
Gene Aliases:
GC79, LGCR
Chromosome Location:
Chr.8: 115408496 - 115669633 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs05523631_s1
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: TRPS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_017013801.1 XP_016869290.1 1 3526 96

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the XM_017013801.1 transcripts.

Set Membership:

Amplicon length between 86 and 100 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Panther Classification:

Molecular Function -

DNA-binding transcription factor

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of transcription from RNA polymerase II promoter
skeletal system development
transcription from RNA polymerase II promoter
NLS-bearing protein import into nucleus
heart development
organ morphogenesis
tissue development
regulation of chondrocyte differentiation
cell fate commitment
positive regulation of transcription from RNA polymerase II promoter
cell development
digestive tract development
anatomical structure formation involved in morphogenesis
RNA polymerase II regulatory region sequence-specific DNA binding
RNA polymerase II transcription factor binding
transcriptional activator activity, RNA polymerase II transcription regulatory region sequence-specific binding
chromatin binding
transcription factor activity, sequence-specific DNA binding
protein binding
zinc ion binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-rr9bx:80/100.66.76.204:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0