This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs05524109_g1
See other SRRM1 GE Assays ›
Gene Symbol:
SRRM1 PRNCR1
Gene Name:
serine and arginine repetitive matrix 1
prostate cancer associated non-coding RNA 1
Gene Aliases:
160-KD, POP101, SRM160
CARLo-3, PCAT8
Chromosome Location:
Chr.1: 24643012 - 24673281 on Build GRCh38
Chromosome Location:
Chr.8: 127079874 - 127092595 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
-
Assay ID Hs05524109_g1
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: SRRM1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001303448.1 NP_001290377.1 17 2822 92
NM_001303449.1 NP_001290378.1 17 2751 92
NM_005839.3 NP_005830.2 16 2786 92
XM_005245717.2 XP_005245774.1 17 2738 92
XM_005245718.2 XP_005245775.1 17 2735 92
XM_005245720.2 XP_005245777.1 17 2729 92
XM_005245721.2 XP_005245778.1 16 2690 92
XM_005245722.2 XP_005245779.1 17 2648 92
XM_011540478.2 XP_011538780.1 17 2752 92
XM_011540479.1 XP_011538781.1 17 4699 92
XM_011540480.1 XP_011538782.1 18 4007 92
XM_011540481.1 XP_011538783.1 18 3049 92
XM_011540482.1 XP_011538784.1 17 4693 92
XM_011540483.1 XP_011538785.1 17 4609 92
XM_017000011.1 XP_016855500.1 17 2749 92
XM_017000012.1 XP_016855501.1 17 2743 92
XM_017000013.1 XP_016855502.1 16 2710 92
XM_017000014.1 XP_016855503.1 16 2707 92
XM_017000015.1 XP_016855504.1 16 2704 92
XM_017000016.1 XP_016855505.1 17 2684 92
XM_017000017.1 XP_016855506.1 17 2659 92
XM_017000018.1 XP_016855507.1 16 2693 92
XM_017000019.1 XP_016855508.1 16 2687 92
XM_017000020.1 XP_016855509.1 17 2645 92
XM_017000021.1 XP_016855510.1 17 2642 92
XM_017000022.1 XP_016855511.1 16 2600 92
XM_017000023.1 XP_016855512.1 17 4696 92
XM_017000024.1 XP_016855513.1 17 4690 92
XM_017000025.1 XP_016855514.1 16 4476 92
XM_017000026.1 XP_016855515.1 17 2826 92
XM_017000027.1 XP_016855516.1 16 4654 92
XM_017000028.1 XP_016855517.1 16 4651 92
XM_017000029.1 XP_016855518.1 17 3001 92
XM_017000030.1 XP_016855519.1 17 4606 92
XM_017000031.1 XP_016855520.1 18 2956 92
XM_017000032.1 XP_016855521.1 17 4603 92
XM_017000033.1 XP_016855522.1 16 4567 92
GenBank mRNA AF048977.1 - 16 2562 92
AF419855.1 - 16 2784 92
AK302498.1 - 17 2641 92
BC036187.1 - 16 2614 4827738 92
KU178512.1 - 16 2562 92
KU178513.1 - 17 2604 92
Gene Symbol: PRNCR1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_109833.1 1 6516 92
GenBank mRNA AB458446.1 - 1 6516 92

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Inventoried Amplicon spans exons and probe does not span exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene SRRM1
Click here to find siRNAs for Gene PRNCR1

Panther Classification:

Molecular Function -

RNA metabolism protein RNA processing factor nucleic acid metabolism protein

Gene Ontology Categories:

Function(s) Process(es)

RNA splicing, via transesterification reactions
mRNA splicing, via spliceosome
termination of RNA polymerase II transcription
RNA export from nucleus
mRNA export from nucleus
RNA splicing
mRNA 3'-end processing
DNA binding
protein binding
poly(A) RNA binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-vsz97:80/100.66.76.49:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0