This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs05530617_s1
See other PLPP4 GE Assays ›
Gene Symbol:
PLPP4
Gene Name:
phospholipid phosphatase 4
Gene Aliases:
DPPL2, PPAPDC1, PPAPDC1A
Chromosome Location:
Chr.10: 120456954 - 120600123 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs05530617_s1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PLPP4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_005269592.2 XP_005269649.1 6 6847 99
XM_006717685.3 XP_006717748.1 7 6687 99
XM_011539444.2 XP_011537746.1 8 6745 99
XM_017015821.1 XP_016871310.1 4 6428 99
XM_017015822.1 XP_016871311.1 4 6395 99

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Panther Classification:

Molecular Function -

phosphatase

Gene Ontology Categories:

Function(s) Process(es)

phospholipid metabolic process
signal transduction
Fc-gamma receptor signaling pathway involved in phagocytosis
phospholipid dephosphorylation
phosphatidate phosphatase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mt7zw:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0